Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151733812-151734355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL2
Entrez Gene ID
344752
Gene Name
arylacetamide deacetylase like 2
Gene Aliases
-
UniGene
Hs.144710
Gene Chromosome Location
Chr.3: 151733916-151758016 on Build GRCh38
Transcripts
NM_207365.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3588856 c.51C>T p.I17I Chr.3: 151734152-151734152 on Build GRCh38
3588854 c.27G>A p.W9* Chr.3: 151734128-151734128 on Build GRCh38
216623 c.40T>A p.L14M Chr.3: 151734141-151734141 on Build GRCh38
1040042 c.42G>T p.L14F Chr.3: 151734143-151734143 on Build GRCh38
728687 c.63T>C p.C21C Chr.3: 151734164-151734164 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777570196 Chr.3: 151734110-151734110 on Build GRCh38
rs774036762 Chr.3: 151734188-151734188 on Build GRCh38
rs769165298 Chr.3: 151734000-151734000 on Build GRCh38
rs537609464 Chr.3: 151734001-151734001 on Build GRCh38
rs1036623026 Chr.3: 151734163-151734163 on Build GRCh38
rs752157269 Chr.3: 151734199-151734199 on Build GRCh38
rs767498542 Chr.3: 151734085-151734085 on Build GRCh38
rs753633453 Chr.3: 151734115-151734115 on Build GRCh38
rs535350087 Chr.3: 151733847-151733847 on Build GRCh38
rs767016169 Chr.3: 151734197-151734197 on Build GRCh38
rs752185946 Chr.3: 151733875-151733875 on Build GRCh38
rs755990497 Chr.3: 151734099-151734099 on Build GRCh38
rs146278742 Chr.3: 151734152-151734152 on Build GRCh38
rs373863706 Chr.3: 151734076-151734076 on Build GRCh38
rs778219200 Chr.3: 151733982-151733982 on Build GRCh38
rs1046383324 Chr.3: 151733879-151733879 on Build GRCh38
rs867406065 Chr.3: 151734166-151734166 on Build GRCh38
rs745993979 Chr.3: 151734140-151734140 on Build GRCh38
rs113990328 Chr.3: 151733976-151733976 on Build GRCh38
rs745933693 Chr.3: 151733987-151733987 on Build GRCh38
rs1055086287 Chr.3: 151734229-151734229 on Build GRCh38
rs757464185 Chr.3: 151734120-151734120 on Build GRCh38
rs374681192 Chr.3: 151734182-151734182 on Build GRCh38
rs555331324 Chr.3: 151734139-151734139 on Build GRCh38
rs1051270038 Chr.3: 151734088-151734088 on Build GRCh38
rs201094017 Chr.3: 151734118-151734118 on Build GRCh38
rs916627996 Chr.3: 151733857-151733857 on Build GRCh38
rs752623427 Chr.3: 151734087-151734087 on Build GRCh38
rs1036882636 Chr.3: 151734138-151734138 on Build GRCh38
rs777077092 Chr.3: 151734028-151734028 on Build GRCh38
rs578230745 Chr.3: 151733968-151733968 on Build GRCh38
rs1025239251 Chr.3: 151734322-151734322 on Build GRCh38
rs924612710 Chr.3: 151733936-151733936 on Build GRCh38
rs558448502 Chr.3: 151733852-151733852 on Build GRCh38
rs201924225 Chr.3: 151734173-151734173 on Build GRCh38
rs566288247 Chr.3: 151733845-151733845 on Build GRCh38
rs971487928 Chr.3: 151733927-151733927 on Build GRCh38
rs912291244 Chr.3: 151734153-151734153 on Build GRCh38
rs542304998 Chr.3: 151734303-151734303 on Build GRCh38
rs377647189 Chr.3: 151734015-151734015 on Build GRCh38
rs1024415797 Chr.3: 151733887-151733887 on Build GRCh38
rs977388329 Chr.3: 151733934-151733934 on Build GRCh38
rs779174108 Chr.3: 151734124-151734124 on Build GRCh38
rs777070007 Chr.3: 151734143-151734143 on Build GRCh38
rs747531513 Chr.3: 151734161-151734161 on Build GRCh38
rs894961041 Chr.3: 151734291-151734294 on Build GRCh38
rs886464667 Chr.3: 151733962-151733962 on Build GRCh38
rs762266308 Chr.3: 151734160-151734160 on Build GRCh38
rs772755346 Chr.3: 151733998-151733998 on Build GRCh38
rs766014988 Chr.3: 151734040-151734040 on Build GRCh38
rs769148557 Chr.3: 151734022-151734022 on Build GRCh38
rs778894507 Chr.3: 151733986-151733986 on Build GRCh38
rs762533216 Chr.3: 151734037-151734037 on Build GRCh38
rs770535804 Chr.3: 151734187-151734187 on Build GRCh38
rs556814703 Chr.3: 151734172-151734172 on Build GRCh38
rs1032559017 Chr.3: 151734317-151734317 on Build GRCh38
rs966755272 Chr.3: 151734234-151734234 on Build GRCh38
rs763991763 Chr.3: 151734210-151734210 on Build GRCh38
rs762206898 Chr.3: 151734180-151734180 on Build GRCh38
rs761190700 Chr.3: 151734019-151734019 on Build GRCh38
rs371094260 Chr.3: 151734034-151734034 on Build GRCh38
rs757949774 Chr.3: 151733953-151733953 on Build GRCh38
rs573659907 Chr.3: 151734208-151734208 on Build GRCh38
rs759110685 Chr.3: 151734190-151734190 on Build GRCh38
rs757063668 Chr.3: 151734223-151734223 on Build GRCh38
rs765506916 Chr.3: 151734028-151734038 on Build GRCh38
rs773290287 Chr.3: 151734196-151734196 on Build GRCh38

View Full Product Details