Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBFB
Entrez Gene ID
865
Gene Name
core-binding factor, beta subunit
Gene Aliases
PEBP2B
Chromosome Location
Chr.16: 67029147 - 67101058 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001755.2 NP_001746.1 4 - 5 662 63
NM_022845.2 NP_074036.1 4 - 5 662 63
XM_006721321.2 XP_006721384.1 3 - 4 507 63
XM_017023812.1 XP_016879301.1 3 - 4 507 63
GenBank mRNA AF294326.1 - 4 - 5 401 63
AK290462.1 - 4 - 5 420 63
AK291834.1 - 4 - 5 404 63
BC018509.1 - 4 - 5 662 63
BM679848.1 - 2 - 3 63
BT006862.1 - 4 - 5 401 63
L20298.1 - 4 - 5 380 63

View Full Product Details