The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001755.2 | NP_001746.1 | 5 - 6 | 787 | 78 |
XM_017023812.1 | XP_016879301.1 | 4 - 5 | 632 | 78 | |
XM_017023813.1 | XP_016879302.1 | 4 - 5 | 656 | 78 | |
GenBank mRNA | AK290462.1 | - | 5 - 6 | 545 | 78 |
AK311084.1 | - | 1 - 2 | 114 | 78 | |
BC018509.1 | - | 5 - 6 | 787 | 78 | |
L20298.1 | - | 5 - 6 | 505 | 78 |