The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002371.3 | NP_002362.1 | 2 - 3 | 400 | 82 |
GenBank mRNA | AK311844.1 | - | 2 - 3 | 347 | 82 |
BC000458.2 | - | 2 - 3 | 371 | 82 | |
BC003006.1 | - | 2 - 3 | 371 | 82 | |
CR541879.1 | - | 2 - 3 | 262 | 82 | |
DT215613.1 | - | 2 - 3 | 403 | 82 | |
M15800.1 | - | 2 - 3 | 316 | 82 | |
X76678.1 | - | 2 - 3 | 262 | 82 |