The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317748.1 | NP_001304677.1 | 15 - 16 | 2750 | 123 |
NM_002380.4 | NP_002371.3 | 16 - 17 | 2873 | 123 | |
XM_005250920.1 | XP_005250977.1 | 15 - 16 | 2424 | 123 | |
GenBank mRNA | AL137638.1 | - | 14 - 15 | 1935 | 123 |
AL833931.1 | - | 16 - 17 | 2761 | 123 | |
AY358895.1 | - | 15 - 16 | 2660 | 123 | |
BC016394.1 | - | 13 - 14 | 1858 | 123 | |
U69263.2 | - | 15 - 16 | 2705 | 123 |