The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032409.2 | NP_001027581.1 | 2 - 3 | 735 | 82 |
NM_001320151.1 | NP_001307080.1 | 2 - 3 | 735 | 82 | |
NM_002534.3 | NP_002525.2 | 2 - 3 | 735 | 82 | |
NM_016816.3 | NP_058132.2 | 2 - 3 | 735 | 82 | |
XM_006719434.1 | XP_006719497.1 | 2 - 3 | 562 | 82 | |
GenBank mRNA | AJ629455.1 | - | 2 - 3 | 505 | 82 |
AK223006.1 | - | 2 - 3 | 82 | ||
AK291003.1 | - | 2 - 3 | 82 | ||
AK300346.1 | - | 2 - 3 | 82 | ||
AY730627.1 | - | 2 - 3 | 82 | ||
AY730628.1 | - | 2 - 3 | 82 | ||
BC000562.2 | - | 2 - 3 | 82 | ||
BC061587.1 | - | 2 - 3 | 503 | 82 | |
BC071981.1 | - | 2 - 3 | 82 | ||
BT006785.1 | - | 2 - 3 | 82 | ||
D00068.1 | - | 2 - 3 | 82 | ||
X02874.1 | - | 2 - 3 | 82 | ||
X04371.1 | - | 2 - 3 | 82 |