The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002890.2 | NP_002881.1 | 9 - 10 | 1531 | 136 |
NM_022650.2 | NP_072179.1 | 9 - 10 | 925 | 136 | |
XM_011543525.1 | XP_011541827.1 | 9 - 10 | 1983 | 136 | |
XM_011543527.2 | XP_011541829.1 | 9 - 10 | 1983 | 136 | |
GenBank mRNA | AB209106.1 | - | 9 - 10 | 1531 | 136 |
AK300263.1 | - | 10 - 11 | 1032 | 136 | |
AK300269.1 | - | 9 - 10 | 858 | 136 | |
AK300406.1 | - | 10 - 11 | 951 | 136 | |
AK312739.1 | - | 9 - 10 | 910 | 136 | |
BC020761.1 | - | 9 - 10 | 912 | 136 | |
BC033015.2 | - | 9 - 10 | 1519 | 136 | |
BC054891.1 | - | 9 - 10 | 891 | 136 | |
BC107870.1 | - | 9 - 10 | 911 | 136 | |
CR749722.1 | - | 9 - 10 | 1896 | 136 | |
M23379.1 | - | 9 - 10 | 1450 | 136 | |
M23612.1 | - | 9 - 10 | 900 | 136 |