Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179576338-179576881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL6A
Entrez Gene ID
86
Gene Name
actin like 6A
Gene Aliases
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K
UniGene
Hs.435326
Gene Chromosome Location
Chr.3: 179562880-179588405 on Build GRCh38
Transcripts
NM_004301.4 NM_177989.3 NM_178042.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3205856 c.733C>G p.Q245E Chr.3: 179576878-179576878 on Build GRCh38
5917955 c.572-5C>T p.? Chr.3: 179576615-179576615 on Build GRCh38
5026083 c.733C>T p.Q245* Chr.3: 179576878-179576878 on Build GRCh38
7317012 c.620G>A p.R207K Chr.3: 179576668-179576668 on Build GRCh38
1617229 c.629T>G p.F210C Chr.3: 179576677-179576677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs117402685 Chr.3: 179576477-179576477 on Build GRCh38
rs373167296 Chr.3: 179576737-179576737 on Build GRCh38
rs374596182 Chr.3: 179576398-179576398 on Build GRCh38
rs528238715 Chr.3: 179576547-179576547 on Build GRCh38
rs977511143 Chr.3: 179576488-179576488 on Build GRCh38
rs34630517 Chr.3: 179576721-179576721 on Build GRCh38
rs369988047 Chr.3: 179576612-179576612 on Build GRCh38
rs878943667 Chr.3: 179576771-179576771 on Build GRCh38
rs563546818 Chr.3: 179576431-179576431 on Build GRCh38
rs746137886 Chr.3: 179576846-179576846 on Build GRCh38
rs748595483 Chr.3: 179576652-179576652 on Build GRCh38
rs778963657 Chr.3: 179576708-179576708 on Build GRCh38
rs763249770 Chr.3: 179576554-179576554 on Build GRCh38
rs751341025 Chr.3: 179576817-179576818 on Build GRCh38
rs1035631360 Chr.3: 179576824-179576824 on Build GRCh38
rs756664674 Chr.3: 179576805-179576805 on Build GRCh38
rs202173390 Chr.3: 179576791-179576791 on Build GRCh38
rs985100935 Chr.3: 179576510-179576510 on Build GRCh38
rs756679524 Chr.3: 179576452-179576452 on Build GRCh38
rs555771748 Chr.3: 179576794-179576794 on Build GRCh38
rs939221385 Chr.3: 179576565-179576565 on Build GRCh38
rs754405685 Chr.3: 179576616-179576616 on Build GRCh38
rs149451650 Chr.3: 179576700-179576700 on Build GRCh38
rs188349278 Chr.3: 179576590-179576590 on Build GRCh38
rs759098849 Chr.3: 179576793-179576793 on Build GRCh38
rs200004281 Chr.3: 179576707-179576707 on Build GRCh38
rs974377910 Chr.3: 179576558-179576558 on Build GRCh38
rs749632307 Chr.3: 179576697-179576697 on Build GRCh38
rs765186027 Chr.3: 179576583-179576583 on Build GRCh38
rs765922265 Chr.3: 179576592-179576592 on Build GRCh38
rs775701995 Chr.3: 179576735-179576735 on Build GRCh38
rs879022012 Chr.3: 179576770-179576770 on Build GRCh38
rs967782198 Chr.3: 179576487-179576487 on Build GRCh38
rs960074927 Chr.3: 179576492-179576492 on Build GRCh38
rs916511838 Chr.3: 179576713-179576713 on Build GRCh38
rs755155418 Chr.3: 179576731-179576731 on Build GRCh38
rs1012231031 Chr.3: 179576461-179576461 on Build GRCh38
rs745968297 Chr.3: 179576712-179576712 on Build GRCh38
rs370585441 Chr.3: 179576746-179576746 on Build GRCh38
rs768017323 Chr.3: 179576761-179576761 on Build GRCh38
rs997708202 Chr.3: 179576842-179576842 on Build GRCh38
rs375773279 Chr.3: 179576829-179576829 on Build GRCh38
rs948024538 Chr.3: 179576553-179576553 on Build GRCh38
rs761718089 Chr.3: 179576578-179576578 on Build GRCh38
rs772337037 Chr.3: 179576848-179576848 on Build GRCh38
rs376110770 Chr.3: 179576738-179576738 on Build GRCh38
rs778205206 Chr.3: 179576813-179576813 on Build GRCh38
rs750241985 Chr.3: 179576588-179576588 on Build GRCh38
rs1046728790 Chr.3: 179576567-179576567 on Build GRCh38
rs74497104 Chr.3: 179576544-179576544 on Build GRCh38
rs898140508 Chr.3: 179576385-179576385 on Build GRCh38
rs773187060 Chr.3: 179576742-179576742 on Build GRCh38
rs758637180 Chr.3: 179576608-179576609 on Build GRCh38
rs34857700 Chr.3: 179576530-179576531 on Build GRCh38
rs539611550 Chr.3: 179576690-179576690 on Build GRCh38
rs1021653338 Chr.3: 179576473-179576473 on Build GRCh38
rs1035745558 Chr.3: 179576489-179576489 on Build GRCh38
rs535487799 Chr.3: 179576781-179576781 on Build GRCh38
rs902040600 Chr.3: 179576365-179576365 on Build GRCh38
rs757357101 Chr.3: 179576423-179576423 on Build GRCh38
rs781724004 Chr.3: 179576617-179576617 on Build GRCh38
rs2292907 Chr.3: 179576564-179576564 on Build GRCh38
rs756597537 Chr.3: 179576676-179576676 on Build GRCh38
rs929763582 Chr.3: 179576386-179576386 on Build GRCh38
rs752198033 Chr.3: 179576795-179576795 on Build GRCh38
rs576520887 Chr.3: 179576432-179576432 on Build GRCh38
rs991971234 Chr.3: 179576521-179576521 on Build GRCh38
rs780193619 Chr.3: 179576759-179576760 on Build GRCh38
rs900097395 Chr.3: 179576453-179576453 on Build GRCh38
rs972542969 Chr.3: 179576555-179576555 on Build GRCh38
rs912314997 Chr.3: 179576568-179576568 on Build GRCh38
rs757498550 Chr.3: 179576833-179576833 on Build GRCh38
rs779212725 Chr.3: 179576838-179576838 on Build GRCh38
rs372804741 Chr.3: 179576782-179576782 on Build GRCh38

View Full Product Details