Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179576746-179577262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL6A
Entrez Gene ID
86
Gene Name
actin like 6A
Gene Aliases
ACTL6, ARPN-BETA, Arp4, BAF53A, INO80K
UniGene
Hs.435326
Gene Chromosome Location
Chr.3: 179562880-179588405 on Build GRCh38
Transcripts
NM_004301.4 NM_177989.3 NM_178042.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3205856 c.733C>G p.Q245E Chr.3: 179576878-179576878 on Build GRCh38
6326387 c.710G>A p.W237* Chr.3: 179576855-179576855 on Build GRCh38
1421065 c.750G>A p.W250* Chr.3: 179576895-179576895 on Build GRCh38
4980397 c.740C>T p.T247M Chr.3: 179576885-179576885 on Build GRCh38
5026083 c.733C>T p.Q245* Chr.3: 179576878-179576878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903464028 Chr.3: 179577129-179577129 on Build GRCh38
rs756870843 Chr.3: 179577234-179577234 on Build GRCh38
rs769871706 Chr.3: 179576886-179576886 on Build GRCh38
rs919843540 Chr.3: 179576968-179576969 on Build GRCh38
rs570980024 Chr.3: 179577190-179577190 on Build GRCh38
rs746137886 Chr.3: 179576846-179576846 on Build GRCh38
rs17838872 Chr.3: 179577207-179577207 on Build GRCh38
rs751341025 Chr.3: 179576817-179576818 on Build GRCh38
rs563585257 Chr.3: 179576990-179576990 on Build GRCh38
rs1035631360 Chr.3: 179576824-179576824 on Build GRCh38
rs1050551336 Chr.3: 179577148-179577148 on Build GRCh38
rs756664674 Chr.3: 179576805-179576805 on Build GRCh38
rs202173390 Chr.3: 179576791-179576791 on Build GRCh38
rs753174247 Chr.3: 179576972-179576972 on Build GRCh38
rs1012992224 Chr.3: 179577176-179577176 on Build GRCh38
rs549919105 Chr.3: 179577208-179577208 on Build GRCh38
rs2292906 Chr.3: 179577013-179577013 on Build GRCh38
rs555771748 Chr.3: 179576794-179576794 on Build GRCh38
rs979373999 Chr.3: 179577054-179577054 on Build GRCh38
rs780324100 Chr.3: 179576862-179576862 on Build GRCh38
rs773240025 Chr.3: 179576896-179576896 on Build GRCh38
rs551080045 Chr.3: 179577188-179577188 on Build GRCh38
rs759098849 Chr.3: 179576793-179576793 on Build GRCh38
rs567246912 Chr.3: 179577186-179577186 on Build GRCh38
rs759176888 Chr.3: 179576936-179576936 on Build GRCh38
rs529228372 Chr.3: 179577004-179577004 on Build GRCh38
rs747059728 Chr.3: 179576885-179576885 on Build GRCh38
rs923389216 Chr.3: 179577141-179577141 on Build GRCh38
rs370130128 Chr.3: 179576946-179576946 on Build GRCh38
rs916459784 Chr.3: 179577038-179577038 on Build GRCh38
rs774180938 Chr.3: 179576926-179576926 on Build GRCh38
rs752228739 Chr.3: 179576944-179576944 on Build GRCh38
rs572346118 Chr.3: 179576913-179576913 on Build GRCh38
rs754289965 Chr.3: 179576960-179576960 on Build GRCh38
rs577935321 Chr.3: 179576969-179576969 on Build GRCh38
rs767178500 Chr.3: 179576937-179576937 on Build GRCh38
rs375773279 Chr.3: 179576829-179576829 on Build GRCh38
rs997708202 Chr.3: 179576842-179576842 on Build GRCh38
rs543597167 Chr.3: 179576982-179576982 on Build GRCh38
rs772337037 Chr.3: 179576848-179576848 on Build GRCh38
rs778205206 Chr.3: 179576813-179576813 on Build GRCh38
rs895802842 Chr.3: 179577153-179577153 on Build GRCh38
rs754763162 Chr.3: 179576922-179576922 on Build GRCh38
rs535487799 Chr.3: 179576781-179576781 on Build GRCh38
rs1037940162 Chr.3: 179577197-179577197 on Build GRCh38
rs752198033 Chr.3: 179576795-179576795 on Build GRCh38
rs868179550 Chr.3: 179576980-179576980 on Build GRCh38
rs933445965 Chr.3: 179577146-179577146 on Build GRCh38
rs2292905 Chr.3: 179577057-179577057 on Build GRCh38
rs559331367 Chr.3: 179577044-179577044 on Build GRCh38
rs781136588 Chr.3: 179576937-179576938 on Build GRCh38
rs778013136 Chr.3: 179577113-179577113 on Build GRCh38
rs762808413 Chr.3: 179576898-179576898 on Build GRCh38
rs1050603697 Chr.3: 179576905-179576905 on Build GRCh38
rs757498550 Chr.3: 179576833-179576833 on Build GRCh38
rs137875597 Chr.3: 179576948-179576948 on Build GRCh38
rs558266605 Chr.3: 179576966-179576966 on Build GRCh38
rs992010798 Chr.3: 179577023-179577023 on Build GRCh38
rs779212725 Chr.3: 179576838-179576838 on Build GRCh38
rs1039859754 Chr.3: 179577079-179577079 on Build GRCh38
rs372804741 Chr.3: 179576782-179576782 on Build GRCh38
rs935366554 Chr.3: 179577106-179577106 on Build GRCh38

View Full Product Details