Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184188677-184189186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCF3
Entrez Gene ID
55324
Gene Name
ATP binding cassette subfamily F member 3
Gene Aliases
EST201864
UniGene
Hs.361323
Gene Chromosome Location
Chr.3: 184186075-184194012 on Build GRCh38
Transcripts
NM_018358.2 XM_011512959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6590665 c.984C>A p.F328L Chr.3: 184189094-184189094 on Build GRCh38
5864018 c.950C>T p.T317I Chr.3: 184188961-184188961 on Build GRCh38
6192834 c.892A>C p.I298L Chr.3: 184188816-184188816 on Build GRCh38
6164131 c.1010C>G p.A337G Chr.3: 184189120-184189120 on Build GRCh38
4630844 c.866C>T p.A289V Chr.3: 184188790-184188790 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772120987 Chr.3: 184189021-184189021 on Build GRCh38
rs762919206 Chr.3: 184188777-184188777 on Build GRCh38
rs755572921 Chr.3: 184189109-184189109 on Build GRCh38
rs1031198210 Chr.3: 184188889-184188889 on Build GRCh38
rs752810860 Chr.3: 184188779-184188779 on Build GRCh38
rs772971982 Chr.3: 184188832-184188832 on Build GRCh38
rs554276240 Chr.3: 184188970-184188970 on Build GRCh38
rs756214841 Chr.3: 184188782-184188782 on Build GRCh38
rs370552813 Chr.3: 184188959-184188959 on Build GRCh38
rs1012414142 Chr.3: 184188817-184188817 on Build GRCh38
rs747708233 Chr.3: 184188912-184188912 on Build GRCh38
rs754073031 Chr.3: 184188875-184188875 on Build GRCh38
rs374622062 Chr.3: 184188988-184188988 on Build GRCh38
rs761914142 Chr.3: 184189031-184189031 on Build GRCh38
rs746520389 Chr.3: 184188800-184188800 on Build GRCh38
rs370370635 Chr.3: 184189043-184189043 on Build GRCh38
rs780539847 Chr.3: 184188895-184188895 on Build GRCh38
rs367733437 Chr.3: 184189072-184189072 on Build GRCh38
rs200129604 Chr.3: 184188722-184188722 on Build GRCh38
rs775032010 Chr.3: 184188743-184188743 on Build GRCh38
rs765043602 Chr.3: 184188767-184188767 on Build GRCh38
rs375473597 Chr.3: 184188925-184188925 on Build GRCh38
rs928065912 Chr.3: 184188860-184188860 on Build GRCh38
rs772174151 Chr.3: 184188946-184188946 on Build GRCh38
rs769424921 Chr.3: 184188916-184188916 on Build GRCh38
rs932085297 Chr.3: 184188730-184188730 on Build GRCh38
rs746547944 Chr.3: 184188757-184188757 on Build GRCh38
rs778732548 Chr.3: 184189005-184189005 on Build GRCh38
rs763543678 Chr.3: 184189125-184189125 on Build GRCh38
rs755345280 Chr.3: 184188700-184188700 on Build GRCh38
rs759962570 Chr.3: 184189081-184189081 on Build GRCh38
rs368097791 Chr.3: 184188944-184188944 on Build GRCh38
rs200656100 Chr.3: 184188729-184188729 on Build GRCh38
rs762039142 Chr.3: 184188950-184188950 on Build GRCh38
rs780652587 Chr.3: 184188911-184188911 on Build GRCh38
rs776829212 Chr.3: 184188949-184188949 on Build GRCh38
rs766675723 Chr.3: 184189063-184189063 on Build GRCh38
rs745689355 Chr.3: 184189033-184189033 on Build GRCh38
rs773979933 Chr.3: 184189155-184189156 on Build GRCh38
rs752763121 Chr.3: 184188720-184188720 on Build GRCh38
rs563979005 Chr.3: 184188754-184188754 on Build GRCh38
rs748007389 Chr.3: 184188865-184188867 on Build GRCh38
rs764290881 Chr.3: 184188871-184188871 on Build GRCh38
rs150245456 Chr.3: 184189128-184189128 on Build GRCh38
rs374209849 Chr.3: 184189046-184189046 on Build GRCh38
rs369759300 Chr.3: 184188735-184188735 on Build GRCh38
rs142912170 Chr.3: 184188987-184188987 on Build GRCh38
rs780942168 Chr.3: 184188895-184188895 on Build GRCh38
rs897099448 Chr.3: 184188771-184188771 on Build GRCh38
rs750784663 Chr.3: 184188884-184188884 on Build GRCh38
rs747832907 Chr.3: 184188827-184188827 on Build GRCh38
rs200941398 Chr.3: 184188900-184188900 on Build GRCh38
rs764346153 Chr.3: 184188786-184188786 on Build GRCh38
rs766278119 Chr.3: 184188778-184188778 on Build GRCh38
rs769478302 Chr.3: 184188831-184188831 on Build GRCh38
rs768337975 Chr.3: 184188762-184188762 on Build GRCh38
rs777916851 Chr.3: 184188723-184188723 on Build GRCh38
rs765601879 Chr.3: 184188881-184188881 on Build GRCh38
rs769790392 Chr.3: 184188877-184188877 on Build GRCh38
rs770490593 Chr.3: 184189050-184189051 on Build GRCh38
rs531703061 Chr.3: 184188790-184188790 on Build GRCh38
rs777471999 Chr.3: 184188917-184188917 on Build GRCh38
rs754126039 Chr.3: 184188787-184188787 on Build GRCh38
rs777342034 Chr.3: 184189088-184189088 on Build GRCh38
rs565912154 Chr.3: 184188945-184188945 on Build GRCh38
rs201803188 Chr.3: 184188826-184188826 on Build GRCh38
rs371486351 Chr.3: 184189129-184189129 on Build GRCh38
rs763058871 Chr.3: 184189048-184189048 on Build GRCh38
rs61745345 Chr.3: 184188799-184188799 on Build GRCh38
rs781709530 Chr.3: 184188973-184188973 on Build GRCh38
rs758269569 Chr.3: 184189153-184189153 on Build GRCh38
rs947950631 Chr.3: 184189152-184189152 on Build GRCh38
rs145869406 Chr.3: 184188764-184188764 on Build GRCh38
rs373441180 Chr.3: 184188856-184188856 on Build GRCh38
rs762094160 Chr.3: 184188879-184188879 on Build GRCh38
rs763316425 Chr.3: 184188961-184188961 on Build GRCh38
rs775632747 Chr.3: 184189022-184189022 on Build GRCh38
rs770718491 Chr.3: 184188994-184188994 on Build GRCh38
rs200981627 Chr.3: 184188717-184188717 on Build GRCh38
rs766338319 Chr.3: 184188841-184188841 on Build GRCh38
rs746272864 Chr.3: 184188797-184188797 on Build GRCh38
rs1023415842 Chr.3: 184188857-184188857 on Build GRCh38
rs757701164 Chr.3: 184188728-184188728 on Build GRCh38
rs376318287 Chr.3: 184189141-184189141 on Build GRCh38
rs896215773 Chr.3: 184188807-184188807 on Build GRCh38
rs138487121 Chr.3: 184188962-184188962 on Build GRCh38
rs376344523 Chr.3: 184188887-184188887 on Build GRCh38
rs753216234 Chr.3: 184189090-184189090 on Build GRCh38
rs767946877 Chr.3: 184189082-184189082 on Build GRCh38
rs765550382 Chr.3: 184188952-184188952 on Build GRCh38
rs778540894 Chr.3: 184188999-184188999 on Build GRCh38
rs751999825 Chr.3: 184188964-184188964 on Build GRCh38
rs843345 Chr.3: 184188727-184188727 on Build GRCh38
rs942008600 Chr.3: 184189006-184189006 on Build GRCh38
rs201987950 Chr.3: 184188897-184188897 on Build GRCh38
rs748797932 Chr.3: 184188897-184188897 on Build GRCh38
rs570723162 Chr.3: 184188805-184188805 on Build GRCh38
rs373175449 Chr.3: 184188738-184188738 on Build GRCh38
rs752200302 Chr.3: 184188985-184188985 on Build GRCh38
rs774160892 Chr.3: 184188851-184188851 on Build GRCh38
rs376213461 Chr.3: 184188763-184188763 on Build GRCh38
rs980082385 Chr.3: 184188709-184188709 on Build GRCh38
rs779450195 Chr.3: 184188793-184188793 on Build GRCh38
rs781093679 Chr.3: 184188859-184188859 on Build GRCh38
rs776220795 Chr.3: 184188815-184188815 on Build GRCh38
rs770088153 Chr.3: 184189027-184189027 on Build GRCh38
rs749046605 Chr.3: 184188930-184188930 on Build GRCh38
rs778684167 Chr.3: 184189137-184189137 on Build GRCh38
rs956964009 Chr.3: 184188893-184188893 on Build GRCh38
rs773096504 Chr.3: 184188772-184188772 on Build GRCh38

View Full Product Details