The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301076.1 | NP_001288005.1 | 2 - 3 | 303 | 63 |
NM_001301081.1 | NP_001288010.1 | 2 - 3 | 260 | 63 | |
NM_004069.4 | NP_004060.2 | 2 - 3 | 378 | 63 | |
GenBank mRNA | AJ010148.1 | - | 1 - 2 | 149 | 63 |
AK312003.1 | - | 2 - 3 | 230 | 63 | |
BC006337.2 | - | 2 - 3 | 187 | 63 | |
BI561502.1 | - | 2 - 3 | 265 | 63 | |
BQ221562.1 | - | 2 - 3 | 303 | 63 | |
CA454481.1 | - | 2 - 3 | 293 | 63 | |
DB472500.1 | - | 2 - 3 | 378 | 63 | |
X97074.1 | - | 2 - 3 | 263 | 63 |