Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3476459-3476953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK7
Entrez Gene ID
285489
Gene Name
docking protein 7
Gene Aliases
C4orf25, CMS10, CMS1B
UniGene
Hs.122110
Gene Chromosome Location
Chr.4: 3463306-3501476 on Build GRCh38
Transcripts
NM_001164673.1 NM_001301071.1 NM_173660.4 XM_011513435.2 XM_011513437.2 XM_017008038.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6167160 c.495C>T p.S165S Chr.4: 3476505-3476505 on Build GRCh38
4550882 c.512G>A p.G171D Chr.4: 3476522-3476522 on Build GRCh38
342483 c.483G>A p.G161G Chr.4: 3476493-3476493 on Build GRCh38
3825775 c.513C>G p.G171G Chr.4: 3476523-3476523 on Build GRCh38
3825776 c.513C>G p.G171G Chr.4: 3476523-3476523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142756576 Chr.4: 3476500-3476500 on Build GRCh38
rs913848578 Chr.4: 3476665-3476665 on Build GRCh38
rs758131044 Chr.4: 3476491-3476491 on Build GRCh38
rs763846570 Chr.4: 3476561-3476561 on Build GRCh38
rs753436594 Chr.4: 3476482-3476482 on Build GRCh38
rs975169509 Chr.4: 3476622-3476622 on Build GRCh38
rs749463997 Chr.4: 3476593-3476593 on Build GRCh38
rs1026589077 Chr.4: 3476597-3476597 on Build GRCh38
rs757138356 Chr.4: 3476569-3476569 on Build GRCh38
rs933831173 Chr.4: 3476640-3476640 on Build GRCh38
rs569830023 Chr.4: 3476521-3476521 on Build GRCh38
rs745897216 Chr.4: 3476589-3476589 on Build GRCh38
rs186594150 Chr.4: 3476738-3476738 on Build GRCh38
rs371609452 Chr.4: 3476496-3476496 on Build GRCh38
rs909622102 Chr.4: 3476615-3476615 on Build GRCh38
rs745956973 Chr.4: 3476508-3476508 on Build GRCh38
rs200487431 Chr.4: 3476560-3476560 on Build GRCh38
rs913807087 Chr.4: 3476841-3476841 on Build GRCh38
rs929461155 Chr.4: 3476904-3476904 on Build GRCh38
rs979769040 Chr.4: 3476858-3476858 on Build GRCh38
rs905198368 Chr.4: 3476677-3476677 on Build GRCh38
rs938066467 Chr.4: 3476689-3476689 on Build GRCh38
rs369531932 Chr.4: 3476690-3476690 on Build GRCh38
rs780241191 Chr.4: 3476592-3476592 on Build GRCh38
rs984602764 Chr.4: 3476694-3476694 on Build GRCh38
rs774633458 Chr.4: 3476582-3476582 on Build GRCh38
rs1018464001 Chr.4: 3476618-3476618 on Build GRCh38
rs771979114 Chr.4: 3476924-3476924 on Build GRCh38
rs896608625 Chr.4: 3476889-3476889 on Build GRCh38
rs767137740 Chr.4: 3476915-3476915 on Build GRCh38
rs116922330 Chr.4: 3476886-3476886 on Build GRCh38
rs150118124 Chr.4: 3476675-3476675 on Build GRCh38
rs946619421 Chr.4: 3476852-3476852 on Build GRCh38
rs574235571 Chr.4: 3476617-3476617 on Build GRCh38
rs1016566702 Chr.4: 3476752-3476752 on Build GRCh38
rs762376200 Chr.4: 3476532-3476532 on Build GRCh38
rs751356491 Chr.4: 3476784-3476784 on Build GRCh38
rs531056781 Chr.4: 3476751-3476751 on Build GRCh38
rs752303492 Chr.4: 3476557-3476557 on Build GRCh38
rs557158726 Chr.4: 3476613-3476613 on Build GRCh38
rs955181083 Chr.4: 3476832-3476832 on Build GRCh38
rs534321926 Chr.4: 3476590-3476590 on Build GRCh38
rs768892432 Chr.4: 3476524-3476524 on Build GRCh38
rs768184265 Chr.4: 3476537-3476537 on Build GRCh38
rs543344523 Chr.4: 3476636-3476636 on Build GRCh38
rs926599835 Chr.4: 3476876-3476876 on Build GRCh38
rs938035233 Chr.4: 3476880-3476880 on Build GRCh38
rs61282869 Chr.4: 3476717-3476717 on Build GRCh38
rs946701004 Chr.4: 3476666-3476666 on Build GRCh38
rs993655277 Chr.4: 3476715-3476715 on Build GRCh38
rs774920743 Chr.4: 3476529-3476529 on Build GRCh38
rs754633490 Chr.4: 3476483-3476483 on Build GRCh38
rs369762997 Chr.4: 3476916-3476916 on Build GRCh38
rs1035116722 Chr.4: 3476586-3476586 on Build GRCh38
rs2699426 Chr.4: 3476596-3476596 on Build GRCh38
rs775583136 Chr.4: 3476523-3476523 on Build GRCh38
rs888073726 Chr.4: 3476729-3476729 on Build GRCh38
rs549987940 Chr.4: 3476808-3476808 on Build GRCh38
rs927164991 Chr.4: 3476746-3476746 on Build GRCh38
rs550024569 Chr.4: 3476490-3476490 on Build GRCh38
rs781227659 Chr.4: 3476506-3476506 on Build GRCh38
rs896639870 Chr.4: 3476693-3476693 on Build GRCh38
rs1056926127 Chr.4: 3476771-3476771 on Build GRCh38
rs2857987 Chr.4: 3476664-3476664 on Build GRCh38
rs60739476 Chr.4: 3476612-3476612 on Build GRCh38
rs746946329 Chr.4: 3476497-3476497 on Build GRCh38
rs550864924 Chr.4: 3476582-3476584 on Build GRCh38
rs374795386 Chr.4: 3476551-3476551 on Build GRCh38
rs371690495 Chr.4: 3476655-3476655 on Build GRCh38
rs963752202 Chr.4: 3476619-3476619 on Build GRCh38
rs922403702 Chr.4: 3476639-3476639 on Build GRCh38
rs781143229 Chr.4: 3476580-3476580 on Build GRCh38
rs563541207 Chr.4: 3476850-3476850 on Build GRCh38
rs751406053 Chr.4: 3476566-3476566 on Build GRCh38
rs1029443704 Chr.4: 3476819-3476819 on Build GRCh38
rs963721238 Chr.4: 3476760-3476760 on Build GRCh38
rs201735755 Chr.4: 3476550-3476550 on Build GRCh38
rs988028205 Chr.4: 3476840-3476840 on Build GRCh38
rs909994359 Chr.4: 3476714-3476714 on Build GRCh38
rs2699427 Chr.4: 3476930-3476930 on Build GRCh38
rs1052367187 Chr.4: 3476494-3476494 on Build GRCh38
rs141667136 Chr.4: 3476603-3476603 on Build GRCh38
rs1047853645 Chr.4: 3476905-3476905 on Build GRCh38
rs368891521 Chr.4: 3476485-3476485 on Build GRCh38
rs1042775367 Chr.4: 3476831-3476831 on Build GRCh38

View Full Product Details