Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXB1
Entrez Gene ID
27023
Gene Name
forkhead box B1
Gene Aliases
FKH5, HFKH-5
Chromosome Location
Chr.15: 60004222 - 60005943 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012182.2 NP_036314.2 2 - 2 963 79
GenBank mRNA AF071554.1 - 1 - 1 508 79
AK290944.1 - 2 - 2 963 79
AK314075.1 - 2 - 2 791 79
BC113710.1 - 1 - 1 546 79

View Full Product Details