Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7758861-7759365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1
Entrez Gene ID
60312
Gene Name
actin filament associated protein 1
Gene Aliases
AFAP, AFAP-110, AFAP110
UniGene
Hs.529369
Gene Chromosome Location
Chr.4: 7758713-7939926 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

Gene Symbol
AFAP1-AS1
Entrez Gene ID
84740
Gene Name
AFAP1 antisense RNA 1
Gene Aliases
AFAP1-AS, AFAP1AS
UniGene
Hs.663029
Gene Chromosome Location
Chr.4: 7754090-7778928 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

SNP Detail

SNP ID Chromosome Location
rs556778443 Chr.4: 7759331-7759331 on Build GRCh38
rs920165184 Chr.4: 7759218-7759218 on Build GRCh38
rs78725170 Chr.4: 7759210-7759210 on Build GRCh38
rs977233259 Chr.4: 7759115-7759115 on Build GRCh38
rs190109880 Chr.4: 7758931-7758931 on Build GRCh38
rs534339756 Chr.4: 7759246-7759246 on Build GRCh38
rs1018431395 Chr.4: 7759195-7759195 on Build GRCh38
rs1004405770 Chr.4: 7759009-7759009 on Build GRCh38
rs115660845 Chr.4: 7759283-7759283 on Build GRCh38
rs879339894 Chr.4: 7759055-7759055 on Build GRCh38
rs564092399 Chr.4: 7759040-7759040 on Build GRCh38
rs776589585 Chr.4: 7758916-7758916 on Build GRCh38
rs562264218 Chr.4: 7759072-7759072 on Build GRCh38
rs942993091 Chr.4: 7758985-7758985 on Build GRCh38
rs1031518425 Chr.4: 7759159-7759159 on Build GRCh38
rs938568187 Chr.4: 7758949-7758949 on Build GRCh38
rs986895634 Chr.4: 7758967-7758967 on Build GRCh38
rs879330180 Chr.4: 7759202-7759202 on Build GRCh38
rs999935928 Chr.4: 7759332-7759332 on Build GRCh38
rs765987060 Chr.4: 7759023-7759023 on Build GRCh38
rs999510839 Chr.4: 7759151-7759151 on Build GRCh38
rs868590075 Chr.4: 7759316-7759316 on Build GRCh38
rs540886241 Chr.4: 7759052-7759052 on Build GRCh38
rs924408069 Chr.4: 7759125-7759125 on Build GRCh38
rs1043385556 Chr.4: 7759324-7759324 on Build GRCh38
rs758242378 Chr.4: 7759020-7759020 on Build GRCh38
rs1048369247 Chr.4: 7759196-7759196 on Build GRCh38
rs1031067362 Chr.4: 7758934-7758934 on Build GRCh38
rs955444495 Chr.4: 7759161-7759161 on Build GRCh38
rs974288342 Chr.4: 7759001-7759001 on Build GRCh38
rs145255135 Chr.4: 7759298-7759298 on Build GRCh38
rs535622716 Chr.4: 7759312-7759312 on Build GRCh38
rs998397556 Chr.4: 7759056-7759056 on Build GRCh38
rs372683455 Chr.4: 7759229-7759229 on Build GRCh38
rs903876957 Chr.4: 7759108-7759108 on Build GRCh38
rs757831811 Chr.4: 7759330-7759330 on Build GRCh38
rs977332400 Chr.4: 7758921-7758921 on Build GRCh38
rs947194571 Chr.4: 7758904-7758904 on Build GRCh38
rs959987032 Chr.4: 7759126-7759126 on Build GRCh38
rs919611323 Chr.4: 7759002-7759002 on Build GRCh38
rs765225770 Chr.4: 7758939-7758939 on Build GRCh38
rs1037203906 Chr.4: 7759268-7759268 on Build GRCh38
rs542382999 Chr.4: 7759081-7759082 on Build GRCh38
rs938504597 Chr.4: 7759054-7759054 on Build GRCh38
rs572264579 Chr.4: 7758935-7758935 on Build GRCh38
rs565667156 Chr.4: 7758994-7758994 on Build GRCh38
rs530673112 Chr.4: 7759191-7759191 on Build GRCh38
rs750208926 Chr.4: 7758984-7758984 on Build GRCh38
rs780693017 Chr.4: 7759139-7759139 on Build GRCh38
rs928378154 Chr.4: 7759282-7759282 on Build GRCh38
rs16841049 Chr.4: 7759099-7759099 on Build GRCh38
rs183134662 Chr.4: 7759083-7759083 on Build GRCh38
rs955748805 Chr.4: 7758945-7758945 on Build GRCh38
rs986278798 Chr.4: 7758886-7758886 on Build GRCh38
rs968473953 Chr.4: 7759114-7759114 on Build GRCh38
rs982433240 Chr.4: 7759242-7759242 on Build GRCh38
rs528475998 Chr.4: 7759044-7759044 on Build GRCh38
rs894313196 Chr.4: 7759090-7759090 on Build GRCh38
rs550926223 Chr.4: 7759098-7759098 on Build GRCh38
rs914444332 Chr.4: 7758887-7758887 on Build GRCh38
rs964167698 Chr.4: 7759208-7759208 on Build GRCh38
rs1021529542 Chr.4: 7759107-7759107 on Build GRCh38
rs894344469 Chr.4: 7759299-7759299 on Build GRCh38
rs986843399 Chr.4: 7759172-7759172 on Build GRCh38
rs894465222 Chr.4: 7758970-7758974 on Build GRCh38

View Full Product Details