Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7762244-7762773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1
Entrez Gene ID
60312
Gene Name
actin filament associated protein 1
Gene Aliases
AFAP, AFAP-110, AFAP110
UniGene
Hs.529369
Gene Chromosome Location
Chr.4: 7758713-7939926 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

Gene Symbol
AFAP1-AS1
Entrez Gene ID
84740
Gene Name
AFAP1 antisense RNA 1
Gene Aliases
AFAP1-AS, AFAP1AS
UniGene
Hs.663029
Gene Chromosome Location
Chr.4: 7754090-7778928 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

SNP Detail

SNP ID Chromosome Location
rs534570345 Chr.4: 7762723-7762723 on Build GRCh38
rs895167961 Chr.4: 7762485-7762485 on Build GRCh38
rs948084433 Chr.4: 7762718-7762718 on Build GRCh38
rs537542897 Chr.4: 7762543-7762543 on Build GRCh38
rs999616517 Chr.4: 7762401-7762401 on Build GRCh38
rs540924947 Chr.4: 7762418-7762418 on Build GRCh38
rs184028157 Chr.4: 7762319-7762319 on Build GRCh38
rs774246923 Chr.4: 7762632-7762632 on Build GRCh38
rs370727462 Chr.4: 7762484-7762484 on Build GRCh38
rs948022170 Chr.4: 7762491-7762491 on Build GRCh38
rs938742185 Chr.4: 7762408-7762408 on Build GRCh38
rs552487390 Chr.4: 7762333-7762333 on Build GRCh38
rs2269849 Chr.4: 7762421-7762421 on Build GRCh38
rs776749340 Chr.4: 7762449-7762450 on Build GRCh38
rs372433619 Chr.4: 7762690-7762690 on Build GRCh38
rs938793208 Chr.4: 7762532-7762532 on Build GRCh38
rs771135583 Chr.4: 7762425-7762425 on Build GRCh38
rs530825599 Chr.4: 7762279-7762279 on Build GRCh38
rs150155041 Chr.4: 7762464-7762464 on Build GRCh38
rs137866246 Chr.4: 7762358-7762358 on Build GRCh38
rs776026033 Chr.4: 7762677-7762677 on Build GRCh38
rs1012356182 Chr.4: 7762325-7762325 on Build GRCh38
rs114444491 Chr.4: 7762494-7762494 on Build GRCh38
rs951144143 Chr.4: 7762656-7762656 on Build GRCh38
rs1048557223 Chr.4: 7762567-7762567 on Build GRCh38
rs992907212 Chr.4: 7762545-7762545 on Build GRCh38
rs938641217 Chr.4: 7762480-7762480 on Build GRCh38
rs763020253 Chr.4: 7762440-7762440 on Build GRCh38
rs1057493795 Chr.4: 7762415-7762415 on Build GRCh38
rs888700722 Chr.4: 7762574-7762574 on Build GRCh38
rs947411676 Chr.4: 7762377-7762377 on Build GRCh38
rs1041690527 Chr.4: 7762390-7762390 on Build GRCh38
rs545822133 Chr.4: 7762397-7762397 on Build GRCh38
rs1046969743 Chr.4: 7762473-7762473 on Build GRCh38
rs993207640 Chr.4: 7762695-7762695 on Build GRCh38
rs79690299 Chr.4: 7762275-7762275 on Build GRCh38
rs563398902 Chr.4: 7762493-7762493 on Build GRCh38
rs1013165624 Chr.4: 7762277-7762277 on Build GRCh38
rs973860660 Chr.4: 7762449-7762449 on Build GRCh38
rs963753995 Chr.4: 7762593-7762593 on Build GRCh38
rs140824807 Chr.4: 7762414-7762414 on Build GRCh38
rs766511029 Chr.4: 7762582-7762582 on Build GRCh38
rs1022778910 Chr.4: 7762327-7762327 on Build GRCh38
rs903858137 Chr.4: 7762732-7762732 on Build GRCh38
rs997265974 Chr.4: 7762469-7762469 on Build GRCh38
rs761929667 Chr.4: 7762646-7762647 on Build GRCh38
rs891268260 Chr.4: 7762530-7762530 on Build GRCh38
rs1031165618 Chr.4: 7762506-7762506 on Build GRCh38
rs764408112 Chr.4: 7762445-7762445 on Build GRCh38
rs929765686 Chr.4: 7762276-7762276 on Build GRCh38
rs932523631 Chr.4: 7762573-7762573 on Build GRCh38
rs907079728 Chr.4: 7762289-7762289 on Build GRCh38
rs10155361 Chr.4: 7762404-7762404 on Build GRCh38
rs1026647760 Chr.4: 7762616-7762616 on Build GRCh38
rs552941885 Chr.4: 7762302-7762302 on Build GRCh38
rs985999535 Chr.4: 7762361-7762361 on Build GRCh38
rs953331266 Chr.4: 7762350-7762350 on Build GRCh38
rs924633036 Chr.4: 7762502-7762502 on Build GRCh38
rs897607729 Chr.4: 7762465-7762465 on Build GRCh38
rs138652108 Chr.4: 7762577-7762577 on Build GRCh38
rs546883317 Chr.4: 7762738-7762738 on Build GRCh38
rs1032112096 Chr.4: 7762679-7762679 on Build GRCh38
rs557051549 Chr.4: 7762343-7762343 on Build GRCh38
rs915974250 Chr.4: 7762556-7762557 on Build GRCh38
rs367736296 Chr.4: 7762323-7762323 on Build GRCh38
rs553944042 Chr.4: 7762648-7762648 on Build GRCh38
rs919640852 Chr.4: 7762450-7762450 on Build GRCh38
rs938490983 Chr.4: 7762681-7762681 on Build GRCh38
rs1030609870 Chr.4: 7762477-7762477 on Build GRCh38
rs968201862 Chr.4: 7762395-7762395 on Build GRCh38
rs928443719 Chr.4: 7762479-7762479 on Build GRCh38
rs529525036 Chr.4: 7762441-7762441 on Build GRCh38
rs570724006 Chr.4: 7762606-7762606 on Build GRCh38
rs914600683 Chr.4: 7762362-7762362 on Build GRCh38
rs1007355682 Chr.4: 7762664-7762664 on Build GRCh38
rs915781007 Chr.4: 7762696-7762696 on Build GRCh38
rs991435665 Chr.4: 7762683-7762683 on Build GRCh38
rs369332033 Chr.4: 7762492-7762492 on Build GRCh38
rs763294391 Chr.4: 7762444-7762444 on Build GRCh38
rs754965341 Chr.4: 7762678-7762678 on Build GRCh38
rs948878180 Chr.4: 7762559-7762560 on Build GRCh38
rs894633919 Chr.4: 7762420-7762420 on Build GRCh38
rs1023334312 Chr.4: 7762284-7762284 on Build GRCh38
rs55794482 Chr.4: 7762290-7762290 on Build GRCh38
rs1056099767 Chr.4: 7762483-7762483 on Build GRCh38
rs977829015 Chr.4: 7762497-7762497 on Build GRCh38
rs888838491 Chr.4: 7762664-7762668 on Build GRCh38

View Full Product Details