Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 7768766-7769271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1
Entrez Gene ID
60312
Gene Name
actin filament associated protein 1
Gene Aliases
AFAP, AFAP-110, AFAP110
UniGene
Hs.529369
Gene Chromosome Location
Chr.4: 7758713-7939926 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

Gene Symbol
AFAP1-AS1
Entrez Gene ID
84740
Gene Name
AFAP1 antisense RNA 1
Gene Aliases
AFAP1-AS, AFAP1AS
UniGene
Hs.663029
Gene Chromosome Location
Chr.4: 7754090-7778928 on Build GRCh38
Transcripts
NM_001134647.1 NM_198595.2 XM_006713909.3 XM_011513544.2 XM_017008535.1 NR_026892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251204 c.2049C>A p.S683R Chr.4: 7768961-7768961 on Build GRCh38
3775993 c.2003C>T p.S668F Chr.4: 7769007-7769007 on Build GRCh38
5401591 c.2160G>A p.K720K Chr.4: 7768850-7768850 on Build GRCh38
5522055 c.2327C>T p.P776L Chr.4: 7768935-7768935 on Build GRCh38
6595737 c.2080C>T p.P694S Chr.4: 7768930-7768930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759126389 Chr.4: 7768989-7768989 on Build GRCh38
rs763909608 Chr.4: 7769189-7769189 on Build GRCh38
rs372536807 Chr.4: 7769022-7769022 on Build GRCh38
rs374522041 Chr.4: 7768859-7768859 on Build GRCh38
rs964302754 Chr.4: 7769210-7769210 on Build GRCh38
rs758053862 Chr.4: 7768995-7768995 on Build GRCh38
rs757167105 Chr.4: 7768939-7768939 on Build GRCh38
rs371528184 Chr.4: 7768887-7768887 on Build GRCh38
rs535933475 Chr.4: 7769087-7769087 on Build GRCh38
rs1044860099 Chr.4: 7769145-7769145 on Build GRCh38
rs552788602 Chr.4: 7769237-7769237 on Build GRCh38
rs1057033722 Chr.4: 7769249-7769249 on Build GRCh38
rs769820127 Chr.4: 7768961-7768961 on Build GRCh38
rs943348508 Chr.4: 7769098-7769098 on Build GRCh38
rs896205206 Chr.4: 7769120-7769120 on Build GRCh38
rs569170923 Chr.4: 7769139-7769139 on Build GRCh38
rs147608397 Chr.4: 7768973-7768973 on Build GRCh38
rs755594567 Chr.4: 7768863-7768863 on Build GRCh38
rs374068705 Chr.4: 7769174-7769174 on Build GRCh38
rs114348397 Chr.4: 7768889-7768889 on Build GRCh38
rs897823763 Chr.4: 7769164-7769164 on Build GRCh38
rs199953617 Chr.4: 7768918-7768918 on Build GRCh38
rs1047898920 Chr.4: 7769143-7769143 on Build GRCh38
rs182900845 Chr.4: 7769250-7769250 on Build GRCh38
rs149064401 Chr.4: 7768990-7768990 on Build GRCh38
rs995052514 Chr.4: 7768835-7768835 on Build GRCh38
rs192944252 Chr.4: 7769112-7769112 on Build GRCh38
rs979149062 Chr.4: 7769079-7769082 on Build GRCh38
rs753365485 Chr.4: 7768927-7768927 on Build GRCh38
rs778959855 Chr.4: 7769024-7769024 on Build GRCh38
rs566902316 Chr.4: 7769209-7769209 on Build GRCh38
rs367769936 Chr.4: 7768943-7768943 on Build GRCh38
rs753441196 Chr.4: 7769192-7769192 on Build GRCh38
rs752516698 Chr.4: 7769160-7769160 on Build GRCh38
rs1007754501 Chr.4: 7769193-7769194 on Build GRCh38
rs930747968 Chr.4: 7769219-7769219 on Build GRCh38
rs968845585 Chr.4: 7769199-7769199 on Build GRCh38
rs567100694 Chr.4: 7768917-7768917 on Build GRCh38
rs947795488 Chr.4: 7769248-7769248 on Build GRCh38
rs530791660 Chr.4: 7769016-7769016 on Build GRCh38
rs974800334 Chr.4: 7769211-7769211 on Build GRCh38
rs748144768 Chr.4: 7768818-7768818 on Build GRCh38
rs1056686346 Chr.4: 7769151-7769151 on Build GRCh38
rs768578290 Chr.4: 7768808-7768808 on Build GRCh38
rs953441130 Chr.4: 7769190-7769190 on Build GRCh38
rs559522095 Chr.4: 7768974-7768974 on Build GRCh38
rs757007611 Chr.4: 7769007-7769007 on Build GRCh38
rs781130898 Chr.4: 7769008-7769008 on Build GRCh38
rs142954070 Chr.4: 7769062-7769062 on Build GRCh38
rs978095251 Chr.4: 7769203-7769203 on Build GRCh38
rs751124922 Chr.4: 7769059-7769059 on Build GRCh38
rs1006567329 Chr.4: 7769156-7769156 on Build GRCh38
rs770107800 Chr.4: 7768901-7768901 on Build GRCh38
rs770979676 Chr.4: 7768907-7768907 on Build GRCh38
rs939603472 Chr.4: 7769148-7769148 on Build GRCh38
rs746578639 Chr.4: 7768836-7768836 on Build GRCh38
rs754653435 Chr.4: 7768876-7768876 on Build GRCh38
rs746819789 Chr.4: 7768865-7768865 on Build GRCh38
rs145262268 Chr.4: 7768933-7768933 on Build GRCh38
rs1056617478 Chr.4: 7769114-7769114 on Build GRCh38
rs778836136 Chr.4: 7768809-7768809 on Build GRCh38
rs768634268 Chr.4: 7768900-7768902 on Build GRCh38
rs535879955 Chr.4: 7768827-7768827 on Build GRCh38
rs983462201 Chr.4: 7769222-7769222 on Build GRCh38
rs749533387 Chr.4: 7769021-7769021 on Build GRCh38
rs1007628758 Chr.4: 7768825-7768825 on Build GRCh38
rs146382774 Chr.4: 7768828-7768828 on Build GRCh38
rs370218957 Chr.4: 7768896-7768896 on Build GRCh38
rs376810371 Chr.4: 7768869-7768869 on Build GRCh38
rs62636596 Chr.4: 7768886-7768886 on Build GRCh38
rs779875998 Chr.4: 7768805-7768805 on Build GRCh38
rs975141470 Chr.4: 7769107-7769107 on Build GRCh38
rs775653718 Chr.4: 7768916-7768916 on Build GRCh38
rs765651163 Chr.4: 7768852-7768852 on Build GRCh38
rs775040596 Chr.4: 7769041-7769041 on Build GRCh38
rs775589151 Chr.4: 7768967-7768967 on Build GRCh38
rs753040534 Chr.4: 7768854-7768854 on Build GRCh38
rs761258356 Chr.4: 7769032-7769032 on Build GRCh38
rs774726162 Chr.4: 7768919-7768919 on Build GRCh38
rs766034305 Chr.4: 7768925-7768925 on Build GRCh38
rs770981973 Chr.4: 7768832-7768832 on Build GRCh38
rs550828358 Chr.4: 7769111-7769111 on Build GRCh38
rs746975098 Chr.4: 7768934-7768934 on Build GRCh38
rs139760292 Chr.4: 7768929-7768929 on Build GRCh38
rs1021787927 Chr.4: 7769194-7769194 on Build GRCh38
rs749461256 Chr.4: 7768970-7768970 on Build GRCh38
rs114583866 Chr.4: 7768994-7768994 on Build GRCh38
rs764771233 Chr.4: 7768995-7768998 on Build GRCh38
rs868729162 Chr.4: 7768935-7768935 on Build GRCh38
rs373992766 Chr.4: 7769033-7769033 on Build GRCh38
rs201575113 Chr.4: 7769027-7769027 on Build GRCh38
rs947922524 Chr.4: 7769132-7769132 on Build GRCh38
rs768168136 Chr.4: 7768975-7768975 on Build GRCh38
rs116834443 Chr.4: 7768913-7768913 on Build GRCh38
rs374242379 Chr.4: 7769015-7769015 on Build GRCh38
rs1043504309 Chr.4: 7768803-7768803 on Build GRCh38
rs762378919 Chr.4: 7768922-7768922 on Build GRCh38
rs977914195 Chr.4: 7768908-7768908 on Build GRCh38
rs772503627 Chr.4: 7769028-7769028 on Build GRCh38
rs369764579 Chr.4: 7769031-7769031 on Build GRCh38
rs1018225506 Chr.4: 7769198-7769198 on Build GRCh38
rs763840650 Chr.4: 7769043-7769043 on Build GRCh38
rs934986484 Chr.4: 7769099-7769100 on Build GRCh38
rs886587535 Chr.4: 7769146-7769146 on Build GRCh38
rs1003186986 Chr.4: 7768870-7768870 on Build GRCh38
rs754496360 Chr.4: 7768928-7768928 on Build GRCh38
rs933875602 Chr.4: 7768915-7768915 on Build GRCh38
rs368807249 Chr.4: 7768874-7768874 on Build GRCh38
rs11933506 Chr.4: 7769236-7769236 on Build GRCh38
rs376932810 Chr.4: 7768806-7768806 on Build GRCh38
rs376912459 Chr.4: 7768976-7768976 on Build GRCh38
rs771872682 Chr.4: 7768824-7768824 on Build GRCh38
rs1056187493 Chr.4: 7768987-7768987 on Build GRCh38
rs563141184 Chr.4: 7769014-7769014 on Build GRCh38
rs139672347 Chr.4: 7768844-7768844 on Build GRCh38
rs776677074 Chr.4: 7768912-7768912 on Build GRCh38
rs776763944 Chr.4: 7768943-7768951 on Build GRCh38
rs540107846 Chr.4: 7769168-7769168 on Build GRCh38
rs1030856027 Chr.4: 7769176-7769176 on Build GRCh38
rs895381889 Chr.4: 7769158-7769158 on Build GRCh38
rs760504602 Chr.4: 7769173-7769173 on Build GRCh38
rs908060144 Chr.4: 7769228-7769228 on Build GRCh38
rs745982783 Chr.4: 7768960-7768960 on Build GRCh38
rs146131879 Chr.4: 7769212-7769212 on Build GRCh38
rs778891105 Chr.4: 7768878-7768878 on Build GRCh38
rs374832060 Chr.4: 7769055-7769055 on Build GRCh38
rs557278704 Chr.4: 7769133-7769133 on Build GRCh38
rs370356365 Chr.4: 7769003-7769003 on Build GRCh38
rs761851060 Chr.4: 7768991-7768992 on Build GRCh38
rs770605051 Chr.4: 7768841-7768841 on Build GRCh38
rs899336966 Chr.4: 7768942-7768942 on Build GRCh38
rs62636595 Chr.4: 7768873-7768873 on Build GRCh38
rs746894354 Chr.4: 7768904-7768904 on Build GRCh38
rs2285761 Chr.4: 7768996-7768996 on Build GRCh38
rs188070339 Chr.4: 7769042-7769042 on Build GRCh38
rs764545264 Chr.4: 7768855-7768855 on Build GRCh38

View Full Product Details