Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
Chromosome Location
Chr.5: 140847772 - 141012344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014005.4 NP_054724.1 1 - 1 5583 109
GenBank mRNA AB002343.1 - 1 - 1 5584 109

View Full Product Details