The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033053.2 | NP_001028225.1 | 6 - 7 | 3256 | 93 |
NM_014922.4 | NP_055737.1 | 6 - 7 | 3256 | 93 | |
NM_033004.3 | NP_127497.1 | 6 - 7 | 3256 | 93 | |
NM_033006.3 | NP_127499.1 | 6 - 7 | 3256 | 93 | |
NM_033007.3 | NP_127500.1 | 6 - 7 | 3256 | 93 | |
GenBank mRNA | AB023143.1 | - | 6 - 7 | 3223 | 93 |
AF229059.1 | - | 6 - 7 | 2701 | 93 | |
AF229060.1 | - | 6 - 7 | 2701 | 93 | |
AF229061.1 | - | 6 - 7 | 2701 | 93 | |
AF229062.1 | - | 6 - 7 | 2701 | 93 | |
AF298548.1 | - | 7 - 8 | 2860 | 93 | |
AF310105.1 | - | 6 - 7 | 3219 | 93 | |
AK026393.1 | - | 3 - 4 | 1179 | 93 | |
AK026398.1 | - | 3 - 4 | 518 | 93 | |
AK310936.1 | - | 7 - 8 | 3067 | 93 | |
AL117470.1 | - | 3 - 4 | 1859 | 93 |