Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr.13: 45954469 - 46052785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015070.4 NP_055885.3 10 - 11 1794 118
XM_005266301.3 XP_005266358.1 10 - 11 2088 118
XM_005266302.3 XP_005266359.1 10 - 11 1788 118
XM_005266303.3 XP_005266360.1 11 - 12 2113 118
XM_005266304.3 XP_005266361.1 10 - 11 2070 118
XM_005266305.3 XP_005266362.1 11 - 12 2106 118
XM_005266306.3 XP_005266363.1 10 - 11 1784 118
XM_005266307.3 XP_005266364.1 10 - 11 2088 118
XM_005266308.3 XP_005266365.1 10 - 11 2088 118
XM_005266309.3 XP_005266366.1 10 - 11 1788 118
XM_005266310.3 XP_005266367.1 10 - 11 2070 118
XM_005266311.3 XP_005266368.1 10 - 11 2088 118
XM_005266312.1 XP_005266369.1 10 - 11 2088 118
XM_017020475.1 XP_016875964.1 11 - 12 1876 118
XM_017020476.1 XP_016875965.1 11 - 12 1873 118
XM_017020477.1 XP_016875966.1 11 - 12 2113 118
XM_017020478.1 XP_016875967.1 11 - 12 2106 118
XM_017020479.1 XP_016875968.1 11 - 12 1876 118
XM_017020480.1 XP_016875969.1 10 - 11 1784 118
XM_017020481.1 XP_016875970.1 11 - 12 2113 118
XM_017020482.1 XP_016875971.1 10 - 11 1788 118
GenBank mRNA AK056422.1 - 10 - 11 2071 118
AK092988.1 - 7 - 8 1433 118
AL136745.1 - 3 - 4 830 118
AL831833.1 - 5 - 6 1198 118
AY283618.1 - 10 - 11 1792 118
BC032425.1 - 3 - 4 962 118
BC050658.2 - 7 - 8 1463 118
BC062685.1 - 3 - 4 860 118
BC107864.1 - 3 - 4 874 118

View Full Product Details