Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 36066976-36067472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP2
Entrez Gene ID
116984
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2
Gene Aliases
CENTD1, PARX
UniGene
Hs.479451
Gene Chromosome Location
Chr.4: 36005186-36244832 on Build GRCh38
Transcripts
NM_015230.3 XM_017007698.1 XM_017007699.1 XM_017007700.1 XM_017007701.1 XM_017007702.1

SNP Detail

SNP ID Chromosome Location
rs1012744677 Chr.4: 36066999-36066999 on Build GRCh38
rs899074299 Chr.4: 36067384-36067384 on Build GRCh38
rs925541353 Chr.4: 36067134-36067134 on Build GRCh38
rs183766578 Chr.4: 36067068-36067068 on Build GRCh38
rs763595876 Chr.4: 36067313-36067313 on Build GRCh38
rs753094691 Chr.4: 36067142-36067144 on Build GRCh38
rs889212239 Chr.4: 36067289-36067289 on Build GRCh38
rs1026494826 Chr.4: 36067063-36067063 on Build GRCh38
rs768038902 Chr.4: 36067023-36067030 on Build GRCh38
rs933494271 Chr.4: 36067239-36067239 on Build GRCh38
rs541105280 Chr.4: 36067177-36067177 on Build GRCh38
rs1051155069 Chr.4: 36067153-36067153 on Build GRCh38
rs992539674 Chr.4: 36067014-36067014 on Build GRCh38
rs978450659 Chr.4: 36067115-36067115 on Build GRCh38
rs984461791 Chr.4: 36067085-36067085 on Build GRCh38
rs548523999 Chr.4: 36067419-36067419 on Build GRCh38
rs914023578 Chr.4: 36067214-36067214 on Build GRCh38
rs907007271 Chr.4: 36067308-36067308 on Build GRCh38
rs893042672 Chr.4: 36067282-36067282 on Build GRCh38
rs1003126845 Chr.4: 36067329-36067329 on Build GRCh38
rs924693952 Chr.4: 36067154-36067154 on Build GRCh38
rs1007496738 Chr.4: 36067301-36067301 on Build GRCh38
rs959908874 Chr.4: 36067008-36067008 on Build GRCh38
rs915684662 Chr.4: 36067018-36067018 on Build GRCh38
rs959835509 Chr.4: 36067412-36067412 on Build GRCh38
rs962961406 Chr.4: 36067116-36067116 on Build GRCh38
rs1005829979 Chr.4: 36067426-36067426 on Build GRCh38
rs559704297 Chr.4: 36067238-36067238 on Build GRCh38
rs975534970 Chr.4: 36067123-36067123 on Build GRCh38
rs993787277 Chr.4: 36067393-36067393 on Build GRCh38
rs1026601141 Chr.4: 36067005-36067005 on Build GRCh38
rs934644599 Chr.4: 36067165-36067165 on Build GRCh38
rs1051774602 Chr.4: 36067274-36067274 on Build GRCh38
rs532519084 Chr.4: 36067048-36067048 on Build GRCh38
rs115602869 Chr.4: 36067408-36067408 on Build GRCh38
rs1052609071 Chr.4: 36067182-36067182 on Build GRCh38
rs936961204 Chr.4: 36067147-36067147 on Build GRCh38
rs145812379 Chr.4: 36067425-36067425 on Build GRCh38
rs950378994 Chr.4: 36067081-36067081 on Build GRCh38
rs762844935 Chr.4: 36067033-36067033 on Build GRCh38
rs934168669 Chr.4: 36067148-36067148 on Build GRCh38

View Full Product Details