The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001251962.1 | NP_001238891.1 | 4 - 5 | 1551 | 76 |
NM_001251963.1 | NP_001238892.1 | 4 - 5 | 1478 | 76 | |
NM_015595.3 | NP_056410.3 | 4 - 5 | 1478 | 76 | |
XM_011512672.1 | XP_011510974.1 | 4 - 5 | 1478 | 76 | |
XM_017006160.1 | XP_016861649.1 | 3 - 4 | 220 | 76 | |
GenBank mRNA | AF415175.1 | - | 4 - 5 | 1433 | 76 |
AK022884.1 | - | 1 - 2 | 65 | 76 | |
AK123040.1 | - | 4 - 5 | 1422 | 76 | |
AK127282.1 | - | 4 - 5 | 1551 | 76 | |
AY552599.1 | - | 3 - 4 | 1267 | 76 | |
BC078655.1 | - | 4 - 5 | 1419 | 76 |