Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41744726-41745231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

SNP Detail

SNP ID Chromosome Location
rs1044665958 Chr.4: 41744966-41744966 on Build GRCh38
rs953745494 Chr.4: 41744793-41744793 on Build GRCh38
rs1053304997 Chr.4: 41745046-41745046 on Build GRCh38
rs973671146 Chr.4: 41744839-41744839 on Build GRCh38
rs992941522 Chr.4: 41744936-41744936 on Build GRCh38
rs919329114 Chr.4: 41744932-41744932 on Build GRCh38
rs549166447 Chr.4: 41744759-41744759 on Build GRCh38
rs958980303 Chr.4: 41745108-41745108 on Build GRCh38
rs932600833 Chr.4: 41745184-41745184 on Build GRCh38
rs902870024 Chr.4: 41744944-41744944 on Build GRCh38
rs371402086 Chr.4: 41744975-41744975 on Build GRCh38
rs776441235 Chr.4: 41744878-41744878 on Build GRCh38
rs899501116 Chr.4: 41745176-41745176 on Build GRCh38
rs886461157 Chr.4: 41745030-41745030 on Build GRCh38
rs977591984 Chr.4: 41745125-41745125 on Build GRCh38
rs962507653 Chr.4: 41744841-41744841 on Build GRCh38
rs973370491 Chr.4: 41745093-41745093 on Build GRCh38
rs868589460 Chr.4: 41745087-41745087 on Build GRCh38
rs573620274 Chr.4: 41745072-41745072 on Build GRCh38
rs189797897 Chr.4: 41744988-41744988 on Build GRCh38
rs752221055 Chr.4: 41744948-41744948 on Build GRCh38
rs924355363 Chr.4: 41745136-41745136 on Build GRCh38
rs1016791592 Chr.4: 41745130-41745130 on Build GRCh38
rs962054269 Chr.4: 41744799-41744799 on Build GRCh38
rs1002992007 Chr.4: 41745001-41745001 on Build GRCh38
rs972567482 Chr.4: 41744845-41744846 on Build GRCh38
rs920820822 Chr.4: 41744845-41744845 on Build GRCh38
rs535962589 Chr.4: 41745114-41745114 on Build GRCh38
rs1029485696 Chr.4: 41744985-41744985 on Build GRCh38
rs73139116 Chr.4: 41745207-41745207 on Build GRCh38
rs993284823 Chr.4: 41745206-41745206 on Build GRCh38
rs867542314 Chr.4: 41744931-41744931 on Build GRCh38
rs551710234 Chr.4: 41744935-41744935 on Build GRCh38
rs745503233 Chr.4: 41745070-41745070 on Build GRCh38
rs575439255 Chr.4: 41745134-41745134 on Build GRCh38
rs1054313192 Chr.4: 41744952-41744952 on Build GRCh38
rs987565912 Chr.4: 41744783-41744783 on Build GRCh38
rs1047746938 Chr.4: 41745013-41745013 on Build GRCh38
rs917417728 Chr.4: 41744916-41744916 on Build GRCh38
rs553814892 Chr.4: 41745027-41745027 on Build GRCh38
rs899725343 Chr.4: 41744963-41744963 on Build GRCh38
rs890950441 Chr.4: 41745000-41745000 on Build GRCh38
rs555491813 Chr.4: 41745120-41745120 on Build GRCh38
rs1027909051 Chr.4: 41745101-41745101 on Build GRCh38
rs544491872 Chr.4: 41745169-41745169 on Build GRCh38
rs759491335 Chr.4: 41744908-41744908 on Build GRCh38
rs991730954 Chr.4: 41745111-41745111 on Build GRCh38
rs184952230 Chr.4: 41744765-41744765 on Build GRCh38
rs1016843942 Chr.4: 41744834-41744834 on Build GRCh38
rs397840867 Chr.4: 41745133-41745134 on Build GRCh38
rs538092014 Chr.4: 41744872-41744872 on Build GRCh38
rs991614725 Chr.4: 41744913-41744913 on Build GRCh38
rs367652707 Chr.4: 41744969-41744969 on Build GRCh38
rs571701837 Chr.4: 41744889-41744889 on Build GRCh38
rs935673383 Chr.4: 41745141-41745141 on Build GRCh38
rs891976504 Chr.4: 41745055-41745055 on Build GRCh38
rs770971831 Chr.4: 41744792-41744792 on Build GRCh38
rs960580004 Chr.4: 41745182-41745182 on Build GRCh38
rs764865376 Chr.4: 41744934-41744934 on Build GRCh38

View Full Product Details