Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41745120-41745614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

SNP Detail

SNP ID Chromosome Location
rs577950819 Chr.4: 41745257-41745257 on Build GRCh38
rs987008229 Chr.4: 41745561-41745561 on Build GRCh38
rs926175692 Chr.4: 41745258-41745258 on Build GRCh38
rs970039324 Chr.4: 41745222-41745222 on Build GRCh38
rs936215651 Chr.4: 41745283-41745283 on Build GRCh38
rs540304746 Chr.4: 41745265-41745265 on Build GRCh38
rs181743762 Chr.4: 41745315-41745315 on Build GRCh38
rs970155283 Chr.4: 41745493-41745493 on Build GRCh38
rs932600833 Chr.4: 41745184-41745184 on Build GRCh38
rs989465234 Chr.4: 41745541-41745541 on Build GRCh38
rs899501116 Chr.4: 41745176-41745176 on Build GRCh38
rs772688984 Chr.4: 41745367-41745367 on Build GRCh38
rs912449909 Chr.4: 41745533-41745533 on Build GRCh38
rs1037976000 Chr.4: 41745390-41745390 on Build GRCh38
rs562632456 Chr.4: 41745434-41745434 on Build GRCh38
rs1051277937 Chr.4: 41745335-41745335 on Build GRCh38
rs898312252 Chr.4: 41745582-41745582 on Build GRCh38
rs763708823 Chr.4: 41745518-41745518 on Build GRCh38
rs958681143 Chr.4: 41745373-41745373 on Build GRCh38
rs939854567 Chr.4: 41745537-41745537 on Build GRCh38
rs73139116 Chr.4: 41745207-41745207 on Build GRCh38
rs201654270 Chr.4: 41745257-41745258 on Build GRCh38
rs551679079 Chr.4: 41745480-41745480 on Build GRCh38
rs993284823 Chr.4: 41745206-41745206 on Build GRCh38
rs920954240 Chr.4: 41745512-41745512 on Build GRCh38
rs1025565125 Chr.4: 41745465-41745465 on Build GRCh38
rs934137643 Chr.4: 41745555-41745555 on Build GRCh38
rs942350095 Chr.4: 41745360-41745361 on Build GRCh38
rs977582808 Chr.4: 41745475-41745475 on Build GRCh38
rs756881848 Chr.4: 41745245-41745245 on Build GRCh38
rs760365271 Chr.4: 41745438-41745438 on Build GRCh38
rs913518069 Chr.4: 41745546-41745546 on Build GRCh38
rs911406786 Chr.4: 41745582-41745585 on Build GRCh38
rs544491872 Chr.4: 41745169-41745169 on Build GRCh38
rs989084011 Chr.4: 41745289-41745289 on Build GRCh38
rs1027689342 Chr.4: 41745359-41745359 on Build GRCh38
rs971984898 Chr.4: 41745445-41745445 on Build GRCh38
rs750862525 Chr.4: 41745298-41745298 on Build GRCh38
rs543147656 Chr.4: 41745421-41745421 on Build GRCh38
rs112714631 Chr.4: 41745542-41745542 on Build GRCh38
rs989845912 Chr.4: 41745489-41745489 on Build GRCh38
rs1015432304 Chr.4: 41745288-41745288 on Build GRCh38
rs913350793 Chr.4: 41745296-41745296 on Build GRCh38
rs371928986 Chr.4: 41745368-41745369 on Build GRCh38
rs73810341 Chr.4: 41745529-41745529 on Build GRCh38
rs577899103 Chr.4: 41745444-41745444 on Build GRCh38
rs957152597 Chr.4: 41745481-41745481 on Build GRCh38
rs1036904128 Chr.4: 41745573-41745573 on Build GRCh38
rs935673383 Chr.4: 41745141-41745141 on Build GRCh38
rs560552240 Chr.4: 41745304-41745304 on Build GRCh38
rs994918200 Chr.4: 41745339-41745339 on Build GRCh38
rs960580004 Chr.4: 41745182-41745182 on Build GRCh38

View Full Product Details