The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016333.3 | NP_057417.3 | 12 - 13 | 8571 | 113 |
XM_005255226.1 | XP_005255283.1 | 12 - 13 | 8549 | 113 | |
GenBank mRNA | AB002322.3 | - | 12 - 13 | 8223 | 113 |
AB016091.1 | - | 2 - 3 | 3593 | 113 | |
AB016092.1 | - | 12 - 13 | 8247 | 113 | |
AF201422.1 | - | 13 - 14 | 6995 | 113 | |
AK025579.1 | - | 2 - 3 | 1696 | 113 | |
AK025903.1 | - | 2 - 3 | 830 | 113 | |
BC009062.1 | - | 2 - 3 | 361 | 113 | |
BC028054.1 | - | 2 - 3 | 2059 | 113 |