Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56338612-56339111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AASDH
Entrez Gene ID
132949
Gene Name
aminoadipate-semialdehyde dehydrogenase
Gene Aliases
ACSF4, LYS2, NRPS1098, NRPS998
UniGene
Hs.104347
Gene Chromosome Location
Chr.4: 56337687-56387677 on Build GRCh38
Transcripts
NM_001286668.1 NM_001286669.1 NM_001286670.1 NM_001286671.1 NM_001323890.1 NM_001323892.1 NM_001323893.1 NM_001323899.1 NM_181806.3 XM_017007740.1 XM_017007741.1 XM_017007742.1 XM_017007743.1 XM_017007744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589324 c.3072G>A p.P1024P Chr.4: 56338627-56338627 on Build GRCh38
3604640 c.2983G>A p.G995S Chr.4: 56338716-56338716 on Build GRCh38
380581 c.2964A>G p.S988S Chr.4: 56338735-56338735 on Build GRCh38
6589321 c.2949G>A p.P983P Chr.4: 56338750-56338750 on Build GRCh38
6512224 c.3055A>T p.R1019W Chr.4: 56338644-56338644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773171531 Chr.4: 56338647-56338647 on Build GRCh38
rs938911775 Chr.4: 56338883-56338884 on Build GRCh38
rs1048622849 Chr.4: 56338925-56338925 on Build GRCh38
rs774612856 Chr.4: 56338694-56338694 on Build GRCh38
rs558367321 Chr.4: 56338856-56338856 on Build GRCh38
rs928368638 Chr.4: 56339084-56339084 on Build GRCh38
rs777947426 Chr.4: 56338758-56338759 on Build GRCh38
rs747192853 Chr.4: 56338815-56338816 on Build GRCh38
rs751190789 Chr.4: 56338688-56338689 on Build GRCh38
rs746009238 Chr.4: 56338801-56338802 on Build GRCh38
rs754596367 Chr.4: 56338728-56338728 on Build GRCh38
rs756598776 Chr.4: 56338722-56338722 on Build GRCh38
rs750385588 Chr.4: 56338661-56338661 on Build GRCh38
rs1017561472 Chr.4: 56338859-56338859 on Build GRCh38
rs1004387142 Chr.4: 56338948-56338949 on Build GRCh38
rs955965609 Chr.4: 56338971-56338971 on Build GRCh38
rs535887461 Chr.4: 56338691-56338691 on Build GRCh38
rs758198512 Chr.4: 56338769-56338769 on Build GRCh38
rs749639479 Chr.4: 56338761-56338762 on Build GRCh38
rs957972105 Chr.4: 56339034-56339034 on Build GRCh38
rs900843913 Chr.4: 56339019-56339019 on Build GRCh38
rs778799153 Chr.4: 56338637-56338637 on Build GRCh38
rs759089734 Chr.4: 56338817-56338817 on Build GRCh38
rs115571683 Chr.4: 56338779-56338779 on Build GRCh38
rs370016579 Chr.4: 56338812-56338813 on Build GRCh38
rs756528818 Chr.4: 56338703-56338703 on Build GRCh38
rs764312880 Chr.4: 56338693-56338693 on Build GRCh38
rs928254066 Chr.4: 56338745-56338745 on Build GRCh38
rs758020310 Chr.4: 56338830-56338830 on Build GRCh38
rs372355217 Chr.4: 56338678-56338678 on Build GRCh38
rs201592398 Chr.4: 56338712-56338712 on Build GRCh38
rs575328567 Chr.4: 56338860-56338860 on Build GRCh38
rs112268987 Chr.4: 56338937-56338937 on Build GRCh38
rs1021440898 Chr.4: 56339030-56339030 on Build GRCh38
rs755947661 Chr.4: 56338686-56338687 on Build GRCh38
rs764889858 Chr.4: 56338820-56338820 on Build GRCh38
rs3733324 Chr.4: 56338794-56338794 on Build GRCh38
rs140104028 Chr.4: 56338774-56338774 on Build GRCh38
rs754465693 Chr.4: 56338816-56338816 on Build GRCh38
rs536834169 Chr.4: 56338828-56338828 on Build GRCh38
rs990408969 Chr.4: 56339024-56339024 on Build GRCh38
rs886833323 Chr.4: 56338937-56338938 on Build GRCh38
rs766142726 Chr.4: 56338658-56338659 on Build GRCh38
rs901890844 Chr.4: 56338889-56338889 on Build GRCh38
rs768519913 Chr.4: 56338817-56338818 on Build GRCh38
rs772084979 Chr.4: 56338807-56338808 on Build GRCh38
rs753414387 Chr.4: 56338725-56338725 on Build GRCh38
rs767871867 Chr.4: 56338797-56338797 on Build GRCh38
rs1057149806 Chr.4: 56338760-56338760 on Build GRCh38
rs544331645 Chr.4: 56338884-56338885 on Build GRCh38
rs938946749 Chr.4: 56338668-56338668 on Build GRCh38
rs774520906 Chr.4: 56338788-56338788 on Build GRCh38
rs1040673378 Chr.4: 56338951-56338951 on Build GRCh38
rs928647123 Chr.4: 56338868-56338868 on Build GRCh38
rs78877748 Chr.4: 56339032-56339032 on Build GRCh38
rs201690440 Chr.4: 56338733-56338733 on Build GRCh38
rs768132543 Chr.4: 56338870-56338870 on Build GRCh38
rs775579900 Chr.4: 56338658-56338658 on Build GRCh38
rs767808606 Chr.4: 56338700-56338700 on Build GRCh38
rs931447806 Chr.4: 56338900-56338900 on Build GRCh38
rs747507318 Chr.4: 56339071-56339072 on Build GRCh38
rs746925401 Chr.4: 56338751-56338751 on Build GRCh38
rs369695347 Chr.4: 56338703-56338703 on Build GRCh38
rs375807933 Chr.4: 56338750-56338750 on Build GRCh38
rs139033817 Chr.4: 56338892-56338892 on Build GRCh38
rs773478279 Chr.4: 56338802-56338802 on Build GRCh38
rs758190607 Chr.4: 56338742-56338742 on Build GRCh38
rs1019709970 Chr.4: 56338770-56338770 on Build GRCh38
rs183689950 Chr.4: 56339033-56339033 on Build GRCh38
rs369772967 Chr.4: 56338652-56338652 on Build GRCh38
rs768789482 Chr.4: 56338783-56338783 on Build GRCh38
rs749354656 Chr.4: 56338780-56338780 on Build GRCh38
rs777559260 Chr.4: 56338835-56338835 on Build GRCh38
rs779391012 Chr.4: 56338795-56338796 on Build GRCh38
rs780861172 Chr.4: 56338941-56338941 on Build GRCh38
rs887875222 Chr.4: 56338798-56338798 on Build GRCh38
rs147527227 Chr.4: 56338763-56338763 on Build GRCh38
rs747800323 Chr.4: 56338739-56338739 on Build GRCh38
rs763093267 Chr.4: 56338685-56338685 on Build GRCh38
rs779224349 Chr.4: 56338776-56338776 on Build GRCh38
rs766982149 Chr.4: 56338809-56338809 on Build GRCh38
rs757742444 Chr.4: 56338765-56338772 on Build GRCh38
rs1007563799 Chr.4: 56338857-56338857 on Build GRCh38
rs1026730991 Chr.4: 56338760-56338761 on Build GRCh38
rs372566623 Chr.4: 56338708-56338708 on Build GRCh38
rs375453713 Chr.4: 56338697-56338697 on Build GRCh38
rs190244294 Chr.4: 56339085-56339085 on Build GRCh38

View Full Product Details