The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282564.1 | NP_001269493.1 | 1 - 2 | 326 | 71 |
NM_018202.5 | NP_060672.2 | 1 - 2 | 326 | 71 | |
XM_005245931.2 | XP_005245988.1 | 1 - 2 | 287 | 71 | |
XM_011541704.2 | XP_011540006.1 | 1 - 2 | 280 | 71 | |
GenBank mRNA | AK001609.1 | - | 1 - 2 | 237 | 71 |
AK311472.1 | - | 1 - 2 | 287 | 71 | |
AY845015.1 | - | 1 - 2 | 256 | 71 | |
AY845035.1 | - | 1 - 2 | 194 | 71 | |
BC032427.1 | - | 1 - 2 | 194 | 71 | |
DB455940.1 | - | 1 - 2 | 326 | 71 |