Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99346815-99347341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH1C
Entrez Gene ID
126
Gene Name
alcohol dehydrogenase 1C (class I), gamma polypeptide
Gene Aliases
ADH3
UniGene
Hs.654537
Gene Chromosome Location
Chr.4: 99336492-99353045 on Build GRCh38
Transcripts
NM_000669.4 NR_133005.1 XM_011531588.2 XM_011531589.1

SNP Detail

SNP ID Chromosome Location
rs113185020 Chr.4: 99347300-99347300 on Build GRCh38
rs148796137 Chr.4: 99346897-99346897 on Build GRCh38
rs952259990 Chr.4: 99346918-99346918 on Build GRCh38
rs951997655 Chr.4: 99346952-99346952 on Build GRCh38
rs774073201 Chr.4: 99346998-99346998 on Build GRCh38
rs1041737842 Chr.4: 99346880-99346880 on Build GRCh38
rs759025388 Chr.4: 99347123-99347123 on Build GRCh38
rs574903402 Chr.4: 99346862-99346862 on Build GRCh38
rs1028708927 Chr.4: 99347259-99347259 on Build GRCh38
rs371594597 Chr.4: 99347039-99347039 on Build GRCh38
rs749381405 Chr.4: 99347100-99347100 on Build GRCh38
rs769953475 Chr.4: 99346986-99346986 on Build GRCh38
rs283413 Chr.4: 99347033-99347033 on Build GRCh38
rs879104375 Chr.4: 99347145-99347146 on Build GRCh38
rs1021781811 Chr.4: 99347301-99347301 on Build GRCh38
rs375611729 Chr.4: 99347189-99347189 on Build GRCh38
rs953493124 Chr.4: 99347013-99347013 on Build GRCh38
rs757743167 Chr.4: 99347105-99347105 on Build GRCh38
rs374241840 Chr.4: 99347082-99347082 on Build GRCh38
rs914530843 Chr.4: 99347014-99347014 on Build GRCh38
rs552443099 Chr.4: 99347168-99347168 on Build GRCh38
rs758612030 Chr.4: 99347278-99347278 on Build GRCh38
rs764059660 Chr.4: 99346995-99346995 on Build GRCh38
rs750038735 Chr.4: 99347035-99347035 on Build GRCh38
rs752060970 Chr.4: 99347135-99347135 on Build GRCh38
rs1014580782 Chr.4: 99346973-99346973 on Build GRCh38
rs1004787403 Chr.4: 99347221-99347221 on Build GRCh38
rs565220194 Chr.4: 99346919-99346919 on Build GRCh38
rs764508784 Chr.4: 99347125-99347125 on Build GRCh38
rs370444853 Chr.4: 99347163-99347163 on Build GRCh38
rs1693431 Chr.4: 99346892-99346892 on Build GRCh38
rs775589950 Chr.4: 99346991-99346991 on Build GRCh38
rs753360532 Chr.4: 99347045-99347045 on Build GRCh38
rs750783783 Chr.4: 99347157-99347157 on Build GRCh38
rs767140954 Chr.4: 99346898-99346898 on Build GRCh38
rs35385902 Chr.4: 99347122-99347122 on Build GRCh38
rs993628790 Chr.4: 99347106-99347106 on Build GRCh38
rs997192899 Chr.4: 99347258-99347258 on Build GRCh38
rs112515298 Chr.4: 99346845-99346845 on Build GRCh38
rs760363342 Chr.4: 99347121-99347121 on Build GRCh38
rs772973805 Chr.4: 99347112-99347112 on Build GRCh38
rs746038367 Chr.4: 99347062-99347062 on Build GRCh38
rs761544047 Chr.4: 99347108-99347108 on Build GRCh38
rs1012113976 Chr.4: 99346944-99346944 on Build GRCh38
rs901502049 Chr.4: 99347241-99347241 on Build GRCh38
rs796094433 Chr.4: 99347006-99347006 on Build GRCh38
rs993596202 Chr.4: 99346893-99346893 on Build GRCh38
rs751880475 Chr.4: 99347051-99347051 on Build GRCh38
rs771806673 Chr.4: 99347109-99347109 on Build GRCh38
rs1019553562 Chr.4: 99346870-99346870 on Build GRCh38
rs372339655 Chr.4: 99346933-99346933 on Build GRCh38
rs151296609 Chr.4: 99347165-99347165 on Build GRCh38
rs890777654 Chr.4: 99347034-99347034 on Build GRCh38
rs1014891087 Chr.4: 99347235-99347235 on Build GRCh38
rs990695208 Chr.4: 99347292-99347292 on Build GRCh38
rs190503441 Chr.4: 99346877-99346877 on Build GRCh38
rs34379379 Chr.4: 99347147-99347147 on Build GRCh38
rs928620699 Chr.4: 99347111-99347111 on Build GRCh38
rs201365141 Chr.4: 99347101-99347101 on Build GRCh38
rs571169213 Chr.4: 99347242-99347242 on Build GRCh38
rs768742583 Chr.4: 99347178-99347178 on Build GRCh38
rs33988198 Chr.4: 99346975-99346975 on Build GRCh38
rs3762896 Chr.4: 99346974-99346974 on Build GRCh38
rs879179212 Chr.4: 99347040-99347040 on Build GRCh38
rs770118058 Chr.4: 99347074-99347074 on Build GRCh38
rs544423606 Chr.4: 99346970-99346970 on Build GRCh38
rs945999506 Chr.4: 99346858-99346858 on Build GRCh38
rs757625289 Chr.4: 99347052-99347052 on Build GRCh38
rs778326690 Chr.4: 99347046-99347046 on Build GRCh38
rs372052813 Chr.4: 99347148-99347148 on Build GRCh38
rs879624336 Chr.4: 99346974-99346975 on Build GRCh38
rs760204028 Chr.4: 99347038-99347038 on Build GRCh38
rs914838106 Chr.4: 99346840-99346840 on Build GRCh38
rs375679943 Chr.4: 99346960-99346960 on Build GRCh38
rs761613003 Chr.4: 99347029-99347029 on Build GRCh38
rs778739003 Chr.4: 99347188-99347188 on Build GRCh38
rs932546861 Chr.4: 99347033-99347033 on Build GRCh38
rs1055829359 Chr.4: 99346926-99346926 on Build GRCh38
rs541840193 Chr.4: 99346864-99346864 on Build GRCh38
rs1027949578 Chr.4: 99346947-99346947 on Build GRCh38

View Full Product Details