Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99412519-99413067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADH7
Entrez Gene ID
131
Gene Name
alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
Gene Aliases
ADH4
UniGene
Hs.389
Gene Chromosome Location
Chr.4: 99412261-99435510 on Build GRCh38
Transcripts
NM_000673.4 NM_001166504.1

SNP Detail

SNP ID Chromosome Location
rs958188984 Chr.4: 99412837-99412837 on Build GRCh38
rs113051735 Chr.4: 99412809-99412809 on Build GRCh38
rs1036961167 Chr.4: 99413040-99413040 on Build GRCh38
rs975470931 Chr.4: 99412552-99412552 on Build GRCh38
rs977288599 Chr.4: 99412884-99412884 on Build GRCh38
rs182155942 Chr.4: 99412824-99412824 on Build GRCh38
rs959249570 Chr.4: 99412591-99412591 on Build GRCh38
rs914472486 Chr.4: 99412602-99412602 on Build GRCh38
rs536427172 Chr.4: 99412767-99412767 on Build GRCh38
rs953300183 Chr.4: 99412575-99412575 on Build GRCh38
rs284786 Chr.4: 99412820-99412820 on Build GRCh38
rs938371175 Chr.4: 99412690-99412690 on Build GRCh38
rs887343409 Chr.4: 99412765-99412765 on Build GRCh38
rs542011738 Chr.4: 99412592-99412592 on Build GRCh38
rs372477436 Chr.4: 99412622-99412623 on Build GRCh38
rs932895043 Chr.4: 99412694-99412694 on Build GRCh38
rs940184716 Chr.4: 99412984-99412984 on Build GRCh38
rs977859170 Chr.4: 99412618-99412618 on Build GRCh38
rs1048588909 Chr.4: 99412728-99412728 on Build GRCh38
rs762746433 Chr.4: 99412996-99412996 on Build GRCh38
rs914724446 Chr.4: 99412976-99412976 on Build GRCh38
rs572888032 Chr.4: 99412806-99412806 on Build GRCh38
rs894230103 Chr.4: 99412725-99412725 on Build GRCh38
rs990382856 Chr.4: 99412862-99412862 on Build GRCh38
rs1041284462 Chr.4: 99412994-99412994 on Build GRCh38
rs756788946 Chr.4: 99412771-99412771 on Build GRCh38
rs944055541 Chr.4: 99413034-99413034 on Build GRCh38
rs897209683 Chr.4: 99412788-99412788 on Build GRCh38
rs1028424256 Chr.4: 99412571-99412571 on Build GRCh38
rs1030764594 Chr.4: 99412930-99412930 on Build GRCh38
rs3805331 Chr.4: 99412775-99412775 on Build GRCh38
rs931105028 Chr.4: 99412937-99412937 on Build GRCh38
rs4147554 Chr.4: 99412909-99412909 on Build GRCh38
rs965433876 Chr.4: 99412549-99412549 on Build GRCh38
rs565756797 Chr.4: 99412626-99412629 on Build GRCh38
rs776048967 Chr.4: 99412802-99412802 on Build GRCh38
rs550427652 Chr.4: 99412726-99412726 on Build GRCh38
rs901145180 Chr.4: 99412823-99412823 on Build GRCh38
rs879120904 Chr.4: 99412631-99412631 on Build GRCh38
rs996848012 Chr.4: 99413007-99413007 on Build GRCh38
rs868000078 Chr.4: 99412999-99412999 on Build GRCh38
rs915635483 Chr.4: 99412912-99412912 on Build GRCh38
rs990002395 Chr.4: 99412580-99412580 on Build GRCh38
rs894369 Chr.4: 99412689-99412689 on Build GRCh38
rs895087360 Chr.4: 99412830-99412830 on Build GRCh38
rs990282789 Chr.4: 99412644-99412644 on Build GRCh38
rs537872787 Chr.4: 99412704-99412704 on Build GRCh38
rs190030483 Chr.4: 99412733-99412733 on Build GRCh38
rs552263689 Chr.4: 99412608-99412608 on Build GRCh38
rs891115532 Chr.4: 99412707-99412707 on Build GRCh38
rs1021490075 Chr.4: 99412876-99412876 on Build GRCh38
rs1049020106 Chr.4: 99412941-99412941 on Build GRCh38
rs542406320 Chr.4: 99412966-99412966 on Build GRCh38
rs967684114 Chr.4: 99412882-99412882 on Build GRCh38
rs946019590 Chr.4: 99412607-99412607 on Build GRCh38
rs912593332 Chr.4: 99413009-99413009 on Build GRCh38
rs1028363742 Chr.4: 99413025-99413025 on Build GRCh38
rs553695512 Chr.4: 99412827-99412827 on Build GRCh38
rs112355520 Chr.4: 99412905-99412905 on Build GRCh38
rs544240324 Chr.4: 99412900-99412900 on Build GRCh38

View Full Product Details