Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99863555-99864053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAPP1
Entrez Gene ID
27071
Gene Name
dual adaptor of phosphotyrosine and 3-phosphoinositides 1
Gene Aliases
BAM32
UniGene
Hs.436271
Gene Chromosome Location
Chr.4: 99816824-99872290 on Build GRCh38
Transcripts
NM_001306151.1 NM_014395.2 XM_011531840.2 XM_011531842.1 XM_011531843.2 XM_017008023.1 XM_017008024.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1426089 c.622C>A p.L208I Chr.4: 99863791-99863791 on Build GRCh38
6325103 c.622C>T p.L208L Chr.4: 99863791-99863791 on Build GRCh38
2951516 c.644C>T p.A215V Chr.4: 99863813-99863813 on Build GRCh38
3760310 c.654C>T p.F218F Chr.4: 99863823-99863823 on Build GRCh38
5400218 c.652T>A p.F218I Chr.4: 99863821-99863821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3737484 Chr.4: 99863823-99863823 on Build GRCh38
rs368508199 Chr.4: 99863980-99863980 on Build GRCh38
rs201852232 Chr.4: 99863759-99863759 on Build GRCh38
rs930536298 Chr.4: 99863597-99863597 on Build GRCh38
rs71913814 Chr.4: 99863719-99863719 on Build GRCh38
rs907708176 Chr.4: 99863610-99863610 on Build GRCh38
rs919905541 Chr.4: 99863998-99863998 on Build GRCh38
rs370086366 Chr.4: 99863752-99863752 on Build GRCh38
rs1054500586 Chr.4: 99863678-99863678 on Build GRCh38
rs985384360 Chr.4: 99863634-99863634 on Build GRCh38
rs919954341 Chr.4: 99863592-99863592 on Build GRCh38
rs773815712 Chr.4: 99863879-99863879 on Build GRCh38
rs769693769 Chr.4: 99863830-99863830 on Build GRCh38
rs753089835 Chr.4: 99863774-99863774 on Build GRCh38
rs776527660 Chr.4: 99863829-99863830 on Build GRCh38
rs146032134 Chr.4: 99863785-99863785 on Build GRCh38
rs771443351 Chr.4: 99863891-99863891 on Build GRCh38
rs745681172 Chr.4: 99863721-99863721 on Build GRCh38
rs913653200 Chr.4: 99863726-99863726 on Build GRCh38
rs1040240170 Chr.4: 99863660-99863660 on Build GRCh38
rs754347073 Chr.4: 99863753-99863753 on Build GRCh38
rs1043952547 Chr.4: 99863777-99863777 on Build GRCh38
rs61359294 Chr.4: 99863735-99863736 on Build GRCh38
rs1020531964 Chr.4: 99863786-99863786 on Build GRCh38
rs577479651 Chr.4: 99863781-99863781 on Build GRCh38
rs757229815 Chr.4: 99863787-99863787 on Build GRCh38
rs775230213 Chr.4: 99863724-99863724 on Build GRCh38
rs566479420 Chr.4: 99863615-99863615 on Build GRCh38
rs768177383 Chr.4: 99863733-99863733 on Build GRCh38
rs572435860 Chr.4: 99863971-99863971 on Build GRCh38
rs995958379 Chr.4: 99863677-99863677 on Build GRCh38
rs768356502 Chr.4: 99863869-99863869 on Build GRCh38
rs970293714 Chr.4: 99863676-99863676 on Build GRCh38
rs764058909 Chr.4: 99863736-99863753 on Build GRCh38
rs560348252 Chr.4: 99863959-99863959 on Build GRCh38
rs761159718 Chr.4: 99863748-99863748 on Build GRCh38
rs765737668 Chr.4: 99863764-99863764 on Build GRCh38
rs575042437 Chr.4: 99863864-99863864 on Build GRCh38
rs779877627 Chr.4: 99863860-99863860 on Build GRCh38
rs867913952 Chr.4: 99863780-99863780 on Build GRCh38
rs766921063 Chr.4: 99863749-99863749 on Build GRCh38
rs939042243 Chr.4: 99863720-99863720 on Build GRCh38
rs796947680 Chr.4: 99863715-99863715 on Build GRCh38
rs761406508 Chr.4: 99863887-99863887 on Build GRCh38
rs761369091 Chr.4: 99863879-99863879 on Build GRCh38
rs745769238 Chr.4: 99863824-99863824 on Build GRCh38
rs958822610 Chr.4: 99863586-99863586 on Build GRCh38
rs115100535 Chr.4: 99863633-99863633 on Build GRCh38
rs559231657 Chr.4: 99863723-99863723 on Build GRCh38
rs1047701917 Chr.4: 99863608-99863608 on Build GRCh38
rs541775818 Chr.4: 99863630-99863630 on Build GRCh38
rs900212165 Chr.4: 99863666-99863666 on Build GRCh38
rs974217043 Chr.4: 99863978-99863978 on Build GRCh38
rs974438603 Chr.4: 99863591-99863591 on Build GRCh38
rs750048939 Chr.4: 99863718-99863719 on Build GRCh38
rs542362703 Chr.4: 99863878-99863878 on Build GRCh38
rs1023549521 Chr.4: 99863958-99863958 on Build GRCh38
rs201164049 Chr.4: 99863782-99863782 on Build GRCh38
rs762639725 Chr.4: 99863727-99863727 on Build GRCh38
rs927628083 Chr.4: 99863719-99863719 on Build GRCh38
rs981695085 Chr.4: 99863719-99863724 on Build GRCh38
rs773975434 Chr.4: 99863741-99863741 on Build GRCh38

View Full Product Details