Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 100398063-100398507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMCN
Entrez Gene ID
51705
Gene Name
endomucin
Gene Aliases
EMCN2, MUC14
UniGene
Hs.152913
Gene Chromosome Location
Chr.4: 100395341-100518093 on Build GRCh38
Transcripts
NM_001159694.1 NM_016242.3

SNP Detail

SNP ID Chromosome Location
rs116166504 Chr.4: 100398260-100398260 on Build GRCh38
rs181777484 Chr.4: 100398431-100398431 on Build GRCh38
rs910262228 Chr.4: 100398152-100398152 on Build GRCh38
rs902620620 Chr.4: 100398311-100398311 on Build GRCh38
rs141521403 Chr.4: 100398137-100398137 on Build GRCh38
rs17552409 Chr.4: 100398175-100398175 on Build GRCh38
rs1057136549 Chr.4: 100398325-100398325 on Build GRCh38
rs1047667386 Chr.4: 100398204-100398204 on Build GRCh38
rs575450182 Chr.4: 100398241-100398241 on Build GRCh38
rs375299601 Chr.4: 100398159-100398159 on Build GRCh38
rs922928395 Chr.4: 100398430-100398430 on Build GRCh38
rs1025507 Chr.4: 100398242-100398242 on Build GRCh38
rs541154883 Chr.4: 100398330-100398330 on Build GRCh38
rs929208571 Chr.4: 100398198-100398198 on Build GRCh38
rs751754546 Chr.4: 100398310-100398310 on Build GRCh38
rs1042832944 Chr.4: 100398111-100398111 on Build GRCh38
rs386400960 Chr.4: 100398390-100398391 on Build GRCh38
rs909136239 Chr.4: 100398213-100398213 on Build GRCh38
rs1012429338 Chr.4: 100398169-100398169 on Build GRCh38
rs779464166 Chr.4: 100398473-100398473 on Build GRCh38
rs190154394 Chr.4: 100398160-100398160 on Build GRCh38
rs983069700 Chr.4: 100398223-100398223 on Build GRCh38
rs137894699 Chr.4: 100398298-100398298 on Build GRCh38
rs976352115 Chr.4: 100398253-100398253 on Build GRCh38
rs931905775 Chr.4: 100398278-100398278 on Build GRCh38
rs397701914 Chr.4: 100398410-100398411 on Build GRCh38
rs946734846 Chr.4: 100398300-100398300 on Build GRCh38
rs74410782 Chr.4: 100398170-100398170 on Build GRCh38
rs545293647 Chr.4: 100398426-100398426 on Build GRCh38
rs969377742 Chr.4: 100398349-100398349 on Build GRCh38
rs533389842 Chr.4: 100398351-100398351 on Build GRCh38
rs893789135 Chr.4: 100398090-100398090 on Build GRCh38
rs372444685 Chr.4: 100398224-100398224 on Build GRCh38
rs562508207 Chr.4: 100398383-100398384 on Build GRCh38
rs891831725 Chr.4: 100398328-100398328 on Build GRCh38
rs905938033 Chr.4: 100398266-100398266 on Build GRCh38
rs866289226 Chr.4: 100398180-100398180 on Build GRCh38
rs141557137 Chr.4: 100398345-100398345 on Build GRCh38
rs1018893999 Chr.4: 100398355-100398356 on Build GRCh38
rs964429110 Chr.4: 100398381-100398392 on Build GRCh38
rs761459040 Chr.4: 100398097-100398097 on Build GRCh38
rs1012460050 Chr.4: 100398384-100398384 on Build GRCh38
rs975830007 Chr.4: 100398418-100398418 on Build GRCh38
rs1049773067 Chr.4: 100398089-100398089 on Build GRCh38
rs759598333 Chr.4: 100398381-100398383 on Build GRCh38
rs386400961 Chr.4: 100398399-100398400 on Build GRCh38
rs138625987 Chr.4: 100398130-100398130 on Build GRCh38
rs755323939 Chr.4: 100398214-100398214 on Build GRCh38
rs35561750 Chr.4: 100398380-100398381 on Build GRCh38
rs374699650 Chr.4: 100398454-100398454 on Build GRCh38
rs766665193 Chr.4: 100398287-100398287 on Build GRCh38
rs946787706 Chr.4: 100398098-100398098 on Build GRCh38

View Full Product Details