Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 145645873-145646417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMAA
Entrez Gene ID
166785
Gene Name
methylmalonic aciduria (cobalamin deficiency) cblA type
Gene Aliases
cblA
UniGene
Hs.452864
Gene Chromosome Location
Chr.4: 145619388-145660035 on Build GRCh38
Transcripts
NM_172250.2 XM_011531684.2 XM_011531685.2 XM_011531686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6488843 c.702G>C p.A234A Chr.4: 145646125-145646125 on Build GRCh38
5501602 c.669C>G p.T223T Chr.4: 145646092-145646092 on Build GRCh38
732482 c.658G>C p.V220L Chr.4: 145646081-145646081 on Build GRCh38
5775785 c.703G>C p.G235R Chr.4: 145646126-145646126 on Build GRCh38
5060151 c.701C>T p.A234V Chr.4: 145646124-145646124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773516780 Chr.4: 145646045-145646045 on Build GRCh38
rs760732816 Chr.4: 145646055-145646055 on Build GRCh38
rs377676676 Chr.4: 145645943-145645943 on Build GRCh38
rs760107279 Chr.4: 145645942-145645942 on Build GRCh38
rs923070882 Chr.4: 145646299-145646299 on Build GRCh38
rs920825350 Chr.4: 145646087-145646087 on Build GRCh38
rs776168690 Chr.4: 145646070-145646070 on Build GRCh38
rs199938583 Chr.4: 145645974-145645974 on Build GRCh38
rs781395862 Chr.4: 145646015-145646015 on Build GRCh38
rs370375675 Chr.4: 145646033-145646033 on Build GRCh38
rs200082858 Chr.4: 145645950-145645950 on Build GRCh38
rs752844506 Chr.4: 145646141-145646141 on Build GRCh38
rs150376474 Chr.4: 145646081-145646081 on Build GRCh38
rs975980557 Chr.4: 145646271-145646271 on Build GRCh38
rs908940900 Chr.4: 145646333-145646333 on Build GRCh38
rs866184737 Chr.4: 145646352-145646352 on Build GRCh38
rs371068331 Chr.4: 145646145-145646145 on Build GRCh38
rs750014244 Chr.4: 145646096-145646096 on Build GRCh38
rs866533353 Chr.4: 145646057-145646057 on Build GRCh38
rs1029096863 Chr.4: 145646009-145646009 on Build GRCh38
rs986396752 Chr.4: 145646324-145646324 on Build GRCh38
rs548523205 Chr.4: 145645928-145645928 on Build GRCh38
rs1046495885 Chr.4: 145646218-145646218 on Build GRCh38
rs771832448 Chr.4: 145646052-145646052 on Build GRCh38
rs864309732 Chr.4: 145646015-145646018 on Build GRCh38
rs1010732692 Chr.4: 145645988-145645988 on Build GRCh38
rs571797666 Chr.4: 145646008-145646008 on Build GRCh38
rs779939886 Chr.4: 145646157-145646157 on Build GRCh38
rs140356252 Chr.4: 145646073-145646073 on Build GRCh38
rs956616814 Chr.4: 145646254-145646254 on Build GRCh38
rs116773849 Chr.4: 145646020-145646020 on Build GRCh38
rs768662983 Chr.4: 145646169-145646169 on Build GRCh38
rs138111772 Chr.4: 145646152-145646152 on Build GRCh38
rs77697341 Chr.4: 145645915-145645915 on Build GRCh38
rs374347679 Chr.4: 145646053-145646053 on Build GRCh38
rs941689476 Chr.4: 145646082-145646082 on Build GRCh38
rs768447137 Chr.4: 145646069-145646069 on Build GRCh38
rs181243452 Chr.4: 145646202-145646202 on Build GRCh38
rs771044996 Chr.4: 145646153-145646153 on Build GRCh38
rs550798043 Chr.4: 145646192-145646192 on Build GRCh38
rs746758730 Chr.4: 145646168-145646168 on Build GRCh38
rs762728569 Chr.4: 145646203-145646203 on Build GRCh38
rs1018915498 Chr.4: 145646035-145646035 on Build GRCh38
rs763324433 Chr.4: 145646097-145646097 on Build GRCh38
rs558375073 Chr.4: 145646276-145646276 on Build GRCh38
rs367609164 Chr.4: 145646124-145646124 on Build GRCh38
rs371897312 Chr.4: 145646191-145646191 on Build GRCh38
rs927691278 Chr.4: 145646219-145646219 on Build GRCh38
rs780433125 Chr.4: 145645985-145645985 on Build GRCh38
rs864309730 Chr.4: 145646076-145646076 on Build GRCh38
rs756944341 Chr.4: 145645954-145645954 on Build GRCh38
rs144389160 Chr.4: 145646010-145646010 on Build GRCh38
rs1036460255 Chr.4: 145646099-145646099 on Build GRCh38
rs935276137 Chr.4: 145645962-145645962 on Build GRCh38
rs953249906 Chr.4: 145646309-145646309 on Build GRCh38
rs751861917 Chr.4: 145646118-145646118 on Build GRCh38
rs934973590 Chr.4: 145646223-145646223 on Build GRCh38
rs1052393023 Chr.4: 145646314-145646314 on Build GRCh38
rs781525637 Chr.4: 145646125-145646125 on Build GRCh38
rs104893849 Chr.4: 145646043-145646043 on Build GRCh38
rs745687257 Chr.4: 145646050-145646050 on Build GRCh38
rs750810986 Chr.4: 145645969-145645969 on Build GRCh38
rs1010765251 Chr.4: 145646393-145646393 on Build GRCh38
rs764850345 Chr.4: 145645957-145645957 on Build GRCh38
rs893686757 Chr.4: 145646319-145646319 on Build GRCh38
rs756221585 Chr.4: 145646144-145646144 on Build GRCh38
rs796051993 Chr.4: 145646016-145646019 on Build GRCh38
rs373957421 Chr.4: 145645970-145645970 on Build GRCh38
rs201970426 Chr.4: 145646205-145646205 on Build GRCh38

View Full Product Details