The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007274.3 | NP_009205.3 | 1 - 2 | 516 | 61 |
GenBank mRNA | AB074415.1 | - | 1 - 2 | 111 | 61 |
AB074416.1 | - | 1 - 2 | 111 | 61 | |
AK057168.1 | - | 1 - 2 | 512 | 61 | |
AK289572.1 | - | 1 - 2 | 127 | 61 | |
BC017365.2 | - | 1 - 2 | 465 | 61 | |
BT006888.1 | - | 1 - 2 | 53 | 61 | |
D88894.2 | - | 1 - 2 | 111 | 61 | |
U91316.1 | - | 1 - 2 | 202 | 61 |