Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 155925108-155925612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSO
Entrez Gene ID
1519
Gene Name
cathepsin O
Gene Aliases
CTSO1
UniGene
Hs.75262
Gene Chromosome Location
Chr.4: 155924118-155953896 on Build GRCh38
Transcripts
NM_001334.2

SNP Detail

SNP ID Chromosome Location
rs995011243 Chr.4: 155925455-155925455 on Build GRCh38
rs1042025960 Chr.4: 155925429-155925429 on Build GRCh38
rs991805727 Chr.4: 155925527-155925527 on Build GRCh38
rs62326161 Chr.4: 155925446-155925446 on Build GRCh38
rs375522967 Chr.4: 155925202-155925202 on Build GRCh38
rs72681588 Chr.4: 155925495-155925495 on Build GRCh38
rs545496718 Chr.4: 155925158-155925158 on Build GRCh38
rs966130018 Chr.4: 155925279-155925279 on Build GRCh38
rs998196586 Chr.4: 155925459-155925459 on Build GRCh38
rs916338148 Chr.4: 155925392-155925392 on Build GRCh38
rs181250369 Chr.4: 155925318-155925318 on Build GRCh38
rs563835665 Chr.4: 155925194-155925194 on Build GRCh38
rs906181385 Chr.4: 155925430-155925430 on Build GRCh38
rs554747676 Chr.4: 155925491-155925491 on Build GRCh38
rs749759707 Chr.4: 155925487-155925487 on Build GRCh38
rs945070245 Chr.4: 155925401-155925401 on Build GRCh38
rs750511902 Chr.4: 155925548-155925548 on Build GRCh38
rs766273896 Chr.4: 155925434-155925434 on Build GRCh38
rs573 Chr.4: 155925456-155925456 on Build GRCh38
rs968643341 Chr.4: 155925510-155925510 on Build GRCh38
rs924176146 Chr.4: 155925333-155925333 on Build GRCh38
rs568408490 Chr.4: 155925514-155925514 on Build GRCh38
rs974829912 Chr.4: 155925307-155925307 on Build GRCh38
rs1029788910 Chr.4: 155925181-155925181 on Build GRCh38
rs542481841 Chr.4: 155925525-155925525 on Build GRCh38
rs3839141 Chr.4: 155925200-155925200 on Build GRCh38
rs995504228 Chr.4: 155925140-155925140 on Build GRCh38
rs1019068140 Chr.4: 155925244-155925244 on Build GRCh38
rs965997071 Chr.4: 155925493-155925493 on Build GRCh38
rs565743092 Chr.4: 155925369-155925369 on Build GRCh38
rs542997453 Chr.4: 155925301-155925301 on Build GRCh38
rs915189223 Chr.4: 155925572-155925572 on Build GRCh38
rs954053508 Chr.4: 155925541-155925541 on Build GRCh38
rs1027896725 Chr.4: 155925153-155925153 on Build GRCh38
rs1005108863 Chr.4: 155925465-155925465 on Build GRCh38
rs111236226 Chr.4: 155925133-155925133 on Build GRCh38
rs901755301 Chr.4: 155925457-155925457 on Build GRCh38
rs762001156 Chr.4: 155925526-155925526 on Build GRCh38
rs983433059 Chr.4: 155925195-155925195 on Build GRCh38
rs188468267 Chr.4: 155925312-155925312 on Build GRCh38
rs947734187 Chr.4: 155925536-155925536 on Build GRCh38
rs767286765 Chr.4: 155925463-155925463 on Build GRCh38
rs1029631786 Chr.4: 155925468-155925468 on Build GRCh38
rs1007354775 Chr.4: 155925492-155925492 on Build GRCh38
rs915156395 Chr.4: 155925374-155925374 on Build GRCh38
rs139291865 Chr.4: 155925338-155925338 on Build GRCh38
rs79348881 Chr.4: 155925200-155925200 on Build GRCh38
rs531245486 Chr.4: 155925256-155925256 on Build GRCh38
rs552748182 Chr.4: 155925443-155925443 on Build GRCh38
rs533098741 Chr.4: 155925419-155925419 on Build GRCh38
rs915368806 Chr.4: 155925207-155925207 on Build GRCh38
rs201823338 Chr.4: 155925199-155925199 on Build GRCh38

View Full Product Details