Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174284107-174284616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP44
Entrez Gene ID
80817
Gene Name
centrosomal protein 44
Gene Aliases
KIAA1712, PS1TP3
UniGene
Hs.555989
Gene Chromosome Location
Chr.4: 174283677-174334355 on Build GRCh38
Transcripts
NM_001040157.2 NM_001145314.1 XM_011532285.2 XM_011532286.2 XM_011532287.2 XM_017008654.1 XM_017008655.1 XM_017008656.1 XM_017008657.1 XM_017008658.1 XM_017008659.1 XM_017008660.1 NM_012180.2

Gene Symbol
FBXO8
Entrez Gene ID
26269
Gene Name
F-box protein 8
Gene Aliases
DC10, FBS, FBX8
UniGene
Hs.76917
Gene Chromosome Location
Chr.4: 174236658-174284251 on Build GRCh38
Transcripts
NM_001040157.2 NM_001145314.1 XM_011532285.2 XM_011532286.2 XM_011532287.2 XM_017008654.1 XM_017008655.1 XM_017008656.1 XM_017008657.1 XM_017008658.1 XM_017008659.1 XM_017008660.1 NM_012180.2

SNP Detail

SNP ID Chromosome Location
rs896310289 Chr.4: 174284184-174284184 on Build GRCh38
rs1033769517 Chr.4: 174284236-174284251 on Build GRCh38
rs191770381 Chr.4: 174284273-174284273 on Build GRCh38
rs969553244 Chr.4: 174284393-174284393 on Build GRCh38
rs546548987 Chr.4: 174284564-174284564 on Build GRCh38
rs952417132 Chr.4: 174284185-174284185 on Build GRCh38
rs973556884 Chr.4: 174284147-174284147 on Build GRCh38
rs758012738 Chr.4: 174284187-174284187 on Build GRCh38
rs553351793 Chr.4: 174284294-174284294 on Build GRCh38
rs577901340 Chr.4: 174284345-174284345 on Build GRCh38
rs1031981199 Chr.4: 174284209-174284210 on Build GRCh38
rs549164722 Chr.4: 174284222-174284222 on Build GRCh38
rs952823058 Chr.4: 174284516-174284516 on Build GRCh38
rs544442952 Chr.4: 174284460-174284460 on Build GRCh38
rs533461216 Chr.4: 174284543-174284543 on Build GRCh38
rs992942340 Chr.4: 174284211-174284211 on Build GRCh38
rs757575110 Chr.4: 174284307-174284307 on Build GRCh38
rs978660410 Chr.4: 174284448-174284448 on Build GRCh38
rs962644040 Chr.4: 174284128-174284128 on Build GRCh38
rs937985325 Chr.4: 174284316-174284316 on Build GRCh38
rs183892397 Chr.4: 174284301-174284301 on Build GRCh38
rs944812170 Chr.4: 174284494-174284494 on Build GRCh38
rs564759343 Chr.4: 174284527-174284527 on Build GRCh38
rs937349494 Chr.4: 174284535-174284535 on Build GRCh38
rs1048412318 Chr.4: 174284296-174284296 on Build GRCh38
rs751077313 Chr.4: 174284283-174284283 on Build GRCh38
rs1036343062 Chr.4: 174284510-174284510 on Build GRCh38
rs553111679 Chr.4: 174284407-174284407 on Build GRCh38
rs1042223120 Chr.4: 174284378-174284378 on Build GRCh38
rs938363745 Chr.4: 174284208-174284208 on Build GRCh38
rs926918289 Chr.4: 174284202-174284202 on Build GRCh38
rs995054463 Chr.4: 174284545-174284545 on Build GRCh38
rs911970470 Chr.4: 174284467-174284467 on Build GRCh38
rs913138524 Chr.4: 174284224-174284224 on Build GRCh38
rs529356128 Chr.4: 174284466-174284466 on Build GRCh38
rs566516872 Chr.4: 174284200-174284200 on Build GRCh38
rs1027850016 Chr.4: 174284158-174284158 on Build GRCh38
rs972892517 Chr.4: 174284501-174284501 on Build GRCh38
rs73870251 Chr.4: 174284413-174284413 on Build GRCh38
rs965918349 Chr.4: 174284280-174284280 on Build GRCh38
rs967242807 Chr.4: 174284447-174284447 on Build GRCh38
rs904381003 Chr.4: 174284271-174284271 on Build GRCh38
rs115411345 Chr.4: 174284324-174284324 on Build GRCh38
rs1042850743 Chr.4: 174284252-174284252 on Build GRCh38
rs992806141 Chr.4: 174284553-174284553 on Build GRCh38
rs575429106 Chr.4: 174284444-174284444 on Build GRCh38
rs535566492 Chr.4: 174284206-174284206 on Build GRCh38
rs932036364 Chr.4: 174284282-174284282 on Build GRCh38
rs966412467 Chr.4: 174284192-174284192 on Build GRCh38
rs764253169 Chr.4: 174284149-174284149 on Build GRCh38
rs995106785 Chr.4: 174284386-174284386 on Build GRCh38
rs972840336 Chr.4: 174284281-174284281 on Build GRCh38
rs1027796292 Chr.4: 174284390-174284390 on Build GRCh38
rs779578187 Chr.4: 174284266-174284266 on Build GRCh38
rs1055444842 Chr.4: 174284322-174284322 on Build GRCh38
rs1054992716 Chr.4: 174284176-174284176 on Build GRCh38
rs905482067 Chr.4: 174284591-174284591 on Build GRCh38
rs144562706 Chr.4: 174284365-174284365 on Build GRCh38
rs73870252 Chr.4: 174284415-174284415 on Build GRCh38
rs931370253 Chr.4: 174284285-174284285 on Build GRCh38
rs912911149 Chr.4: 174284445-174284445 on Build GRCh38
rs147850394 Chr.4: 174284441-174284441 on Build GRCh38
rs1050723048 Chr.4: 174284463-174284463 on Build GRCh38
rs35787697 Chr.4: 174284551-174284552 on Build GRCh38
rs569056723 Chr.4: 174284243-174284243 on Build GRCh38
rs529933668 Chr.4: 174284428-174284428 on Build GRCh38
rs931816866 Chr.4: 174284461-174284461 on Build GRCh38
rs984167505 Chr.4: 174284518-174284518 on Build GRCh38
rs945994210 Chr.4: 174284230-174284230 on Build GRCh38

View Full Product Details