Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 174298859-174299371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP44
Entrez Gene ID
80817
Gene Name
centrosomal protein 44
Gene Aliases
KIAA1712, PS1TP3
UniGene
Hs.555989
Gene Chromosome Location
Chr.4: 174283677-174334355 on Build GRCh38
Transcripts
NM_001040157.2 NM_001145314.1 XM_011532285.2 XM_011532286.2 XM_011532287.2 XM_017008654.1 XM_017008655.1 XM_017008656.1 XM_017008657.1 XM_017008658.1 XM_017008659.1 XM_017008660.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6099890 c.32G>A p.R11Q Chr.4: 174299153-174299153 on Build GRCh38
6099889 c.32G>A p.R11Q Chr.4: 174299153-174299153 on Build GRCh38
3974813 c.52C>T p.R18C Chr.4: 174299173-174299173 on Build GRCh38
1053280 c.8C>T p.T3I Chr.4: 174299129-174299129 on Build GRCh38
5692113 c.67C>A p.P23T Chr.4: 174299188-174299188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs889217581 Chr.4: 174298952-174298952 on Build GRCh38
rs917145067 Chr.4: 174298890-174298890 on Build GRCh38
rs549454056 Chr.4: 174298990-174298990 on Build GRCh38
rs746149670 Chr.4: 174299091-174299091 on Build GRCh38
rs1007528902 Chr.4: 174298967-174298967 on Build GRCh38
rs770624346 Chr.4: 174299139-174299139 on Build GRCh38
rs753944057 Chr.4: 174299083-174299083 on Build GRCh38
rs758507613 Chr.4: 174299188-174299188 on Build GRCh38
rs924547215 Chr.4: 174299241-174299241 on Build GRCh38
rs550510000 Chr.4: 174299298-174299298 on Build GRCh38
rs374332972 Chr.4: 174299082-174299082 on Build GRCh38
rs1041533653 Chr.4: 174298898-174298898 on Build GRCh38
rs201923363 Chr.4: 174299072-174299072 on Build GRCh38
rs560523931 Chr.4: 174298948-174298948 on Build GRCh38
rs751849303 Chr.4: 174299197-174299197 on Build GRCh38
rs571940664 Chr.4: 174299136-174299136 on Build GRCh38
rs569255001 Chr.4: 174299018-174299018 on Build GRCh38
rs766900436 Chr.4: 174298924-174298924 on Build GRCh38
rs762459349 Chr.4: 174299137-174299137 on Build GRCh38
rs556185774 Chr.4: 174299331-174299331 on Build GRCh38
rs752157695 Chr.4: 174298991-174298991 on Build GRCh38
rs368228207 Chr.4: 174299185-174299185 on Build GRCh38
rs756875136 Chr.4: 174299222-174299222 on Build GRCh38
rs1041545905 Chr.4: 174299230-174299233 on Build GRCh38
rs777202712 Chr.4: 174299225-174299225 on Build GRCh38
rs768996936 Chr.4: 174299104-174299104 on Build GRCh38
rs759422144 Chr.4: 174299151-174299151 on Build GRCh38
rs950017519 Chr.4: 174298892-174298892 on Build GRCh38
rs754808396 Chr.4: 174299079-174299079 on Build GRCh38
rs1040498965 Chr.4: 174299294-174299294 on Build GRCh38
rs557298077 Chr.4: 174299252-174299252 on Build GRCh38
rs750534448 Chr.4: 174299174-174299174 on Build GRCh38
rs751387997 Chr.4: 174299077-174299077 on Build GRCh38
rs949787067 Chr.4: 174299201-174299201 on Build GRCh38
rs775438472 Chr.4: 174299256-174299256 on Build GRCh38
rs368903445 Chr.4: 174298949-174298949 on Build GRCh38
rs770444038 Chr.4: 174299244-174299244 on Build GRCh38
rs749798126 Chr.4: 174299139-174299141 on Build GRCh38
rs993490094 Chr.4: 174299319-174299319 on Build GRCh38
rs903016686 Chr.4: 174298907-174298907 on Build GRCh38
rs774090871 Chr.4: 174299248-174299248 on Build GRCh38
rs759244784 Chr.4: 174299251-174299251 on Build GRCh38
rs757449216 Chr.4: 174299085-174299085 on Build GRCh38
rs748712246 Chr.4: 174299216-174299216 on Build GRCh38
rs374364731 Chr.4: 174299196-174299196 on Build GRCh38
rs769361456 Chr.4: 174299131-174299131 on Build GRCh38
rs748864789 Chr.4: 174299226-174299226 on Build GRCh38
rs758659662 Chr.4: 174299095-174299095 on Build GRCh38
rs777172405 Chr.4: 174299112-174299112 on Build GRCh38
rs755411530 Chr.4: 174299208-174299208 on Build GRCh38
rs769120665 Chr.4: 174299172-174299183 on Build GRCh38
rs867254400 Chr.4: 174299333-174299333 on Build GRCh38
rs778095178 Chr.4: 174299079-174299080 on Build GRCh38
rs772045617 Chr.4: 174299253-174299253 on Build GRCh38
rs909614262 Chr.4: 174299152-174299152 on Build GRCh38
rs771226342 Chr.4: 174299239-174299241 on Build GRCh38
rs189626839 Chr.4: 174299308-174299308 on Build GRCh38
rs747430320 Chr.4: 174299100-174299100 on Build GRCh38
rs377466253 Chr.4: 174299147-174299147 on Build GRCh38
rs757895917 Chr.4: 174299315-174299315 on Build GRCh38
rs778434529 Chr.4: 174299262-174299262 on Build GRCh38
rs1041643573 Chr.4: 174299240-174299240 on Build GRCh38
rs73011419 Chr.4: 174299153-174299153 on Build GRCh38
rs554199972 Chr.4: 174299157-174299157 on Build GRCh38
rs370516561 Chr.4: 174299166-174299166 on Build GRCh38
rs770304205 Chr.4: 174299223-174299223 on Build GRCh38
rs567241640 Chr.4: 174299273-174299273 on Build GRCh38
rs781663252 Chr.4: 174299214-174299214 on Build GRCh38
rs180885711 Chr.4: 174298943-174298943 on Build GRCh38
rs780214478 Chr.4: 174299097-174299097 on Build GRCh38
rs375288124 Chr.4: 174299173-174299173 on Build GRCh38
rs747626793 Chr.4: 174299113-174299113 on Build GRCh38
rs183743788 Chr.4: 174298970-174298970 on Build GRCh38

View Full Product Details