Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC71
Entrez Gene ID
64925
Gene Name
coiled-coil domain containing 71
Gene Aliases
-
Chromosome Location
Chr.3: 49162535 - 49166352 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK023691.1 - 1 - 1 1256 57

View Full Product Details