Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGFR2
Entrez Gene ID
2263
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location
Chr.10: 121478330 - 121598656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144913.1 NP_001138385.1 7 - 8 1236 120
NM_001144919.1 NP_001138391.1 7 - 8 1466 120
NM_022970.3 NP_075259.4 8 - 9 1733 120
XM_006717708.2 XP_006717771.1 8 - 9 1931 120
XM_006717710.3 XP_006717773.1 8 - 9 1931 120
XM_006717711.2 XP_006717774.1 6 - 7 969 120
XM_006717712.2 XP_006717775.1 5 - 6 891 120
XM_017015920.1 XP_016871409.1 8 - 9 1931 120
XM_017015922.1 XP_016871411.1 6 - 7 969 120
GenBank mRNA AB030073.1 - 8 - 9 1504 120
AB030074.1 - 8 - 9 1504 120
AB030075.1 - 8 - 9 1504 120
AB030076.1 - 8 - 9 1504 120
AB030077.1 - 8 - 9 1504 120
AB030078.1 - 8 - 9 1504 120
AK026508.1 - 7 - 8 1440 120
KP192323.1 - 6 - 7 903 120
KP192324.1 - 6 - 7 903 120
M80634.1 - 8 - 9 1504 120
M97193.1 - 8 - 9 1697 120
X56191.1 - 7 - 8 1110 120

View Full Product Details