Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184763033-184763543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL1
Entrez Gene ID
2180
Gene Name
acyl-CoA synthetase long-chain family member 1
Gene Aliases
ACS1, FACL1, FACL2, LACS, LACS1, LACS2
UniGene
Hs.406678
Gene Chromosome Location
Chr.4: 184755595-184826593 on Build GRCh38
Transcripts
NM_001286708.1 NM_001286710.1 NM_001286711.1 NM_001286712.1 NM_001995.3 XM_005262828.1 XM_005262829.1 XM_005262831.1 XM_011531742.1 XM_017007887.1 XM_017007888.1 XM_017007889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1671454 c.1454C>T p.P485L Chr.4: 184763234-184763234 on Build GRCh38
396560 c.1433G>T p.G478V Chr.4: 184763255-184763255 on Build GRCh38
1428866 c.1448C>T p.P483L Chr.4: 184763240-184763240 on Build GRCh38
6234874 c.1517_1521+1delGCGAGG p.? Chr.4: 184763166-184763171 on Build GRCh38
1428865 c.1513G>A p.E505K Chr.4: 184763175-184763175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768222770 Chr.4: 184763354-184763354 on Build GRCh38
rs923344813 Chr.4: 184763359-184763359 on Build GRCh38
rs1055823993 Chr.4: 184763303-184763303 on Build GRCh38
rs181589674 Chr.4: 184763123-184763123 on Build GRCh38
rs769235113 Chr.4: 184763190-184763190 on Build GRCh38
rs746349597 Chr.4: 184763208-184763208 on Build GRCh38
rs913749377 Chr.4: 184763173-184763173 on Build GRCh38
rs765322754 Chr.4: 184763268-184763268 on Build GRCh38
rs1017288312 Chr.4: 184763507-184763507 on Build GRCh38
rs373958064 Chr.4: 184763233-184763233 on Build GRCh38
rs1045900840 Chr.4: 184763353-184763353 on Build GRCh38
rs755266544 Chr.4: 184763282-184763282 on Build GRCh38
rs201800442 Chr.4: 184763153-184763153 on Build GRCh38
rs201779955 Chr.4: 184763154-184763154 on Build GRCh38
rs965906150 Chr.4: 184763511-184763511 on Build GRCh38
rs41278585 Chr.4: 184763170-184763170 on Build GRCh38
rs375741961 Chr.4: 184763117-184763117 on Build GRCh38
rs779295849 Chr.4: 184763144-184763144 on Build GRCh38
rs953446803 Chr.4: 184763380-184763380 on Build GRCh38
rs1007617972 Chr.4: 184763480-184763480 on Build GRCh38
rs772913726 Chr.4: 184763479-184763479 on Build GRCh38
rs897551312 Chr.4: 184763069-184763069 on Build GRCh38
rs200424498 Chr.4: 184763175-184763175 on Build GRCh38
rs542122098 Chr.4: 184763360-184763360 on Build GRCh38
rs759966643 Chr.4: 184763154-184763154 on Build GRCh38
rs762669593 Chr.4: 184763119-184763119 on Build GRCh38
rs923323148 Chr.4: 184763155-184763155 on Build GRCh38
rs377663790 Chr.4: 184763288-184763288 on Build GRCh38
rs750654477 Chr.4: 184763275-184763275 on Build GRCh38
rs556186174 Chr.4: 184763315-184763315 on Build GRCh38
rs759497640 Chr.4: 184763129-184763129 on Build GRCh38
rs774184895 Chr.4: 184763239-184763239 on Build GRCh38
rs772759019 Chr.4: 184763228-184763228 on Build GRCh38
rs887561290 Chr.4: 184763442-184763442 on Build GRCh38
rs769324831 Chr.4: 184763304-184763304 on Build GRCh38
rs561767010 Chr.4: 184763387-184763387 on Build GRCh38
rs761506246 Chr.4: 184763435-184763435 on Build GRCh38
rs986539074 Chr.4: 184763388-184763388 on Build GRCh38
rs935986855 Chr.4: 184763251-184763251 on Build GRCh38
rs993255792 Chr.4: 184763077-184763077 on Build GRCh38
rs747661349 Chr.4: 184763182-184763182 on Build GRCh38
rs950294965 Chr.4: 184763345-184763345 on Build GRCh38
rs748603684 Chr.4: 184763296-184763296 on Build GRCh38
rs370223408 Chr.4: 184763176-184763176 on Build GRCh38
rs540839724 Chr.4: 184763415-184763415 on Build GRCh38
rs757340868 Chr.4: 184763137-184763137 on Build GRCh38
rs751946654 Chr.4: 184763280-184763280 on Build GRCh38
rs770681248 Chr.4: 184763234-184763234 on Build GRCh38
rs916127690 Chr.4: 184763321-184763321 on Build GRCh38
rs539451395 Chr.4: 184763276-184763276 on Build GRCh38
rs1049277300 Chr.4: 184763433-184763433 on Build GRCh38
rs1052274686 Chr.4: 184763244-184763244 on Build GRCh38
rs966011198 Chr.4: 184763160-184763160 on Build GRCh38
rs111530757 Chr.4: 184763311-184763311 on Build GRCh38
rs984928309 Chr.4: 184763124-184763124 on Build GRCh38
rs576452760 Chr.4: 184763332-184763332 on Build GRCh38
rs535735332 Chr.4: 184763135-184763135 on Build GRCh38
rs762010439 Chr.4: 184763261-184763261 on Build GRCh38
rs1013357962 Chr.4: 184763255-184763255 on Build GRCh38
rs1015065005 Chr.4: 184763347-184763347 on Build GRCh38
rs980404049 Chr.4: 184763106-184763106 on Build GRCh38
rs199793459 Chr.4: 184763245-184763245 on Build GRCh38
rs898000758 Chr.4: 184763391-184763391 on Build GRCh38
rs558474416 Chr.4: 184763200-184763200 on Build GRCh38
rs780338745 Chr.4: 184763277-184763277 on Build GRCh38
rs751482547 Chr.4: 184763128-184763128 on Build GRCh38
rs572219387 Chr.4: 184763405-184763405 on Build GRCh38
rs759613911 Chr.4: 184763242-184763242 on Build GRCh38
rs767110359 Chr.4: 184763472-184763472 on Build GRCh38
rs773820563 Chr.4: 184763404-184763404 on Build GRCh38
rs752627811 Chr.4: 184763249-184763249 on Build GRCh38
rs764329774 Chr.4: 184763130-184763130 on Build GRCh38
rs41278583 Chr.4: 184763093-184763093 on Build GRCh38
rs772853841 Chr.4: 184763118-184763118 on Build GRCh38

View Full Product Details