Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 184764772-184765277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSL1
Entrez Gene ID
2180
Gene Name
acyl-CoA synthetase long-chain family member 1
Gene Aliases
ACS1, FACL1, FACL2, LACS, LACS1, LACS2
UniGene
Hs.406678
Gene Chromosome Location
Chr.4: 184755595-184826593 on Build GRCh38
Transcripts
NM_001286708.1 NM_001286710.1 NM_001286711.1 NM_001286712.1 NM_001995.3 XM_005262828.1 XM_005262829.1 XM_005262831.1 XM_011531742.1 XM_017007887.1 XM_017007888.1 XM_017007889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987833 c.1390A>G p.T464A Chr.4: 184764895-184764895 on Build GRCh38
3993584 c.1427C>T p.T476I Chr.4: 184764858-184764858 on Build GRCh38
4796756 c.1417G>A p.G473R Chr.4: 184764868-184764868 on Build GRCh38
7354023 c.1409C>G p.T470S Chr.4: 184764876-184764876 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943561861 Chr.4: 184764796-184764796 on Build GRCh38
rs763147009 Chr.4: 184764848-184764848 on Build GRCh38
rs964957444 Chr.4: 184765092-184765092 on Build GRCh38
rs753242610 Chr.4: 184764863-184764863 on Build GRCh38
rs763104518 Chr.4: 184764957-184764957 on Build GRCh38
rs747667769 Chr.4: 184765169-184765169 on Build GRCh38
rs756601419 Chr.4: 184764874-184764874 on Build GRCh38
rs573759448 Chr.4: 184765099-184765099 on Build GRCh38
rs1006198611 Chr.4: 184765074-184765074 on Build GRCh38
rs138936282 Chr.4: 184765104-184765104 on Build GRCh38
rs1053965999 Chr.4: 184764820-184764820 on Build GRCh38
rs543187209 Chr.4: 184765083-184765083 on Build GRCh38
rs866671555 Chr.4: 184764882-184764882 on Build GRCh38
rs34311553 Chr.4: 184764983-184764983 on Build GRCh38
rs895125202 Chr.4: 184765197-184765197 on Build GRCh38
rs773325257 Chr.4: 184764841-184764841 on Build GRCh38
rs760662749 Chr.4: 184764836-184764836 on Build GRCh38
rs939374195 Chr.4: 184765249-184765249 on Build GRCh38
rs534607805 Chr.4: 184764995-184764995 on Build GRCh38
rs947912395 Chr.4: 184765156-184765156 on Build GRCh38
rs916238593 Chr.4: 184765149-184765149 on Build GRCh38
rs7653899 Chr.4: 184764977-184764977 on Build GRCh38
rs558138393 Chr.4: 184764948-184764950 on Build GRCh38
rs146216950 Chr.4: 184764857-184764857 on Build GRCh38
rs978261325 Chr.4: 184765118-184765118 on Build GRCh38
rs745481028 Chr.4: 184764895-184764895 on Build GRCh38
rs190785639 Chr.4: 184765091-184765091 on Build GRCh38
rs893955513 Chr.4: 184764828-184764828 on Build GRCh38
rs370966189 Chr.4: 184764856-184764856 on Build GRCh38
rs765740036 Chr.4: 184765126-184765126 on Build GRCh38
rs943614237 Chr.4: 184765078-184765078 on Build GRCh38
rs748951900 Chr.4: 184764806-184764806 on Build GRCh38
rs756648605 Chr.4: 184765101-184765101 on Build GRCh38
rs565542303 Chr.4: 184765235-184765235 on Build GRCh38
rs757390357 Chr.4: 184765055-184765055 on Build GRCh38
rs2292898 Chr.4: 184764878-184764878 on Build GRCh38
rs577433562 Chr.4: 184765072-184765072 on Build GRCh38
rs775602034 Chr.4: 184764832-184764832 on Build GRCh38
rs759419555 Chr.4: 184764819-184764819 on Build GRCh38
rs771947260 Chr.4: 184764829-184764829 on Build GRCh38
rs559484771 Chr.4: 184765105-184765105 on Build GRCh38
rs777304792 Chr.4: 184764805-184764805 on Build GRCh38
rs767829093 Chr.4: 184764858-184764858 on Build GRCh38
rs776363360 Chr.4: 184764940-184764940 on Build GRCh38
rs201691298 Chr.4: 184764837-184764837 on Build GRCh38
rs985144799 Chr.4: 184765028-184765028 on Build GRCh38
rs1047835090 Chr.4: 184765020-184765020 on Build GRCh38
rs947962765 Chr.4: 184764981-184764981 on Build GRCh38
rs376631586 Chr.4: 184764813-184764813 on Build GRCh38
rs7654285 Chr.4: 184765067-184765067 on Build GRCh38
rs961226272 Chr.4: 184765244-184765244 on Build GRCh38
rs922367265 Chr.4: 184764996-184764996 on Build GRCh38
rs34107464 Chr.4: 184764911-184764911 on Build GRCh38
rs775407611 Chr.4: 184764910-184764910 on Build GRCh38
rs774585918 Chr.4: 184764976-184764976 on Build GRCh38
rs7654302 Chr.4: 184765136-184765136 on Build GRCh38
rs756665731 Chr.4: 184764889-184764889 on Build GRCh38
rs112501096 Chr.4: 184765188-184765188 on Build GRCh38
rs1005446671 Chr.4: 184765214-184765214 on Build GRCh38
rs906366085 Chr.4: 184764984-184764984 on Build GRCh38
rs376479044 Chr.4: 184764908-184764908 on Build GRCh38
rs768630298 Chr.4: 184764928-184764928 on Build GRCh38
rs781657529 Chr.4: 184765056-184765056 on Build GRCh38
rs549916725 Chr.4: 184764854-184764854 on Build GRCh38
rs191307498 Chr.4: 184764823-184764823 on Build GRCh38
rs150765294 Chr.4: 184764890-184764890 on Build GRCh38
rs867943863 Chr.4: 184765084-184765084 on Build GRCh38
rs766319165 Chr.4: 184764965-184764965 on Build GRCh38
rs202046765 Chr.4: 184765234-184765234 on Build GRCh38
rs545362641 Chr.4: 184765209-184765209 on Build GRCh38
rs748836543 Chr.4: 184764887-184764887 on Build GRCh38
rs7678719 Chr.4: 184764942-184764942 on Build GRCh38
rs185935681 Chr.4: 184765050-184765050 on Build GRCh38
rs958042637 Chr.4: 184765146-184765146 on Build GRCh38
rs759995634 Chr.4: 184765062-184765062 on Build GRCh38
rs994040331 Chr.4: 184765013-184765013 on Build GRCh38
rs183249574 Chr.4: 184765147-184765147 on Build GRCh38
rs571849237 Chr.4: 184764815-184764815 on Build GRCh38

View Full Product Details