The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023919.2 | NP_076408.1 | 1 - 1 | 897 | 140 |
GenBank mRNA | BC069091.1 | - | 1 - 1 | 840 | 140 |
BC093992.1 | - | 1 - 1 | 897 | 140 | |
BC113561.1 | - | 1 - 1 | 897 | 140 |