Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 225780-226313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHA
Entrez Gene ID
6389
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Gene Aliases
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
UniGene
Hs.440475
Gene Chromosome Location
Chr.5: 218223-264816 on Build GRCh38
Transcripts
NM_001294332.1 NM_004168.3 XM_005248331.3 XM_011514072.1 XM_011514073.1 XM_017009685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7389027 c.589G>T p.G197C Chr.5: 226015-226015 on Build GRCh38
4849570 c.485G>C p.R162T Chr.5: 225911-225911 on Build GRCh38
5809763 c.511C>T p.R171C Chr.5: 225937-225937 on Build GRCh38
6925930 c.460G>A p.E154K Chr.5: 225886-225886 on Build GRCh38
4398703 c.465T>G p.N155K Chr.5: 225891-225891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762379590 Chr.5: 225862-225863 on Build GRCh38
rs148246073 Chr.5: 225976-225976 on Build GRCh38
rs6555056 Chr.5: 226107-226107 on Build GRCh38
rs774589410 Chr.5: 225893-225893 on Build GRCh38
rs777873911 Chr.5: 225887-225887 on Build GRCh38
rs757707693 Chr.5: 225972-225972 on Build GRCh38
rs750177090 Chr.5: 225861-225861 on Build GRCh38
rs908237877 Chr.5: 226215-226215 on Build GRCh38
rs749329370 Chr.5: 225960-225960 on Build GRCh38
rs750444002 Chr.5: 226096-226096 on Build GRCh38
rs188139287 Chr.5: 226199-226199 on Build GRCh38
rs747737117 Chr.5: 226062-226062 on Build GRCh38
rs770052391 Chr.5: 225879-225879 on Build GRCh38
rs989318548 Chr.5: 225894-225894 on Build GRCh38
rs755158523 Chr.5: 226216-226216 on Build GRCh38
rs6555057 Chr.5: 226149-226149 on Build GRCh38
rs764113865 Chr.5: 225841-225841 on Build GRCh38
rs199618059 Chr.5: 225984-225984 on Build GRCh38
rs876658486 Chr.5: 226041-226041 on Build GRCh38
rs61733344 Chr.5: 225975-225975 on Build GRCh38
rs201967413 Chr.5: 226026-226026 on Build GRCh38
rs375067326 Chr.5: 225914-225914 on Build GRCh38
rs760136774 Chr.5: 225834-225834 on Build GRCh38
rs759827541 Chr.5: 225902-225902 on Build GRCh38
rs137982834 Chr.5: 226175-226175 on Build GRCh38
rs773334818 Chr.5: 226000-226000 on Build GRCh38
rs778632786 Chr.5: 225860-225860 on Build GRCh38
rs370853909 Chr.5: 226150-226150 on Build GRCh38
rs1034877127 Chr.5: 226181-226181 on Build GRCh38
rs1011448841 Chr.5: 226136-226136 on Build GRCh38
rs141874250 Chr.5: 226023-226023 on Build GRCh38
rs878854632 Chr.5: 225881-225883 on Build GRCh38
rs748265289 Chr.5: 225872-225872 on Build GRCh38
rs771741537 Chr.5: 225903-225903 on Build GRCh38
rs756351128 Chr.5: 225957-225957 on Build GRCh38
rs776301796 Chr.5: 226061-226061 on Build GRCh38
rs908267345 Chr.5: 225994-225994 on Build GRCh38
rs996865414 Chr.5: 225825-225825 on Build GRCh38
rs371775474 Chr.5: 225867-225867 on Build GRCh38
rs749824479 Chr.5: 225890-225890 on Build GRCh38
rs551900309 Chr.5: 226206-226206 on Build GRCh38
rs181278759 Chr.5: 225942-225942 on Build GRCh38
rs765899092 Chr.5: 225875-225875 on Build GRCh38
rs763578369 Chr.5: 225925-225925 on Build GRCh38
rs535270890 Chr.5: 226079-226079 on Build GRCh38
rs6555055 Chr.5: 226045-226045 on Build GRCh38
rs766258363 Chr.5: 225946-225946 on Build GRCh38
rs765111856 Chr.5: 225999-225999 on Build GRCh38
rs921029131 Chr.5: 226284-226284 on Build GRCh38
rs766339992 Chr.5: 226011-226011 on Build GRCh38
rs750447329 Chr.5: 226190-226190 on Build GRCh38
rs534645492 Chr.5: 226282-226282 on Build GRCh38
rs575770636 Chr.5: 226081-226081 on Build GRCh38
rs753760172 Chr.5: 225845-225845 on Build GRCh38
rs546362356 Chr.5: 226082-226082 on Build GRCh38
rs765358591 Chr.5: 226095-226095 on Build GRCh38
rs753964643 Chr.5: 226191-226191 on Build GRCh38
rs866396717 Chr.5: 226068-226068 on Build GRCh38
rs373340696 Chr.5: 226033-226033 on Build GRCh38
rs775827529 Chr.5: 225979-225979 on Build GRCh38
rs941115978 Chr.5: 226237-226237 on Build GRCh38
rs780411478 Chr.5: 226046-226046 on Build GRCh38
rs769454045 Chr.5: 225954-225954 on Build GRCh38
rs765157205 Chr.5: 225939-225939 on Build GRCh38
rs150646390 Chr.5: 226028-226028 on Build GRCh38
rs772849135 Chr.5: 226066-226066 on Build GRCh38
rs757215259 Chr.5: 225850-225850 on Build GRCh38
rs769390635 Chr.5: 226063-226063 on Build GRCh38
rs185729748 Chr.5: 226165-226165 on Build GRCh38
rs878854633 Chr.5: 226022-226022 on Build GRCh38
rs6555058 Chr.5: 226261-226261 on Build GRCh38
rs751890312 Chr.5: 226099-226099 on Build GRCh38
rs764307917 Chr.5: 226024-226024 on Build GRCh38
rs763046170 Chr.5: 226074-226074 on Build GRCh38
rs386684229 Chr.5: 226279-226282 on Build GRCh38
rs61754481 Chr.5: 226039-226039 on Build GRCh38
rs770937658 Chr.5: 226076-226076 on Build GRCh38
rs377459480 Chr.5: 226097-226097 on Build GRCh38
rs770407681 Chr.5: 226021-226021 on Build GRCh38
rs139881415 Chr.5: 225989-225989 on Build GRCh38
rs762956849 Chr.5: 226010-226010 on Build GRCh38
rs753004001 Chr.5: 225949-225949 on Build GRCh38
rs751451425 Chr.5: 226174-226174 on Build GRCh38
rs528514697 Chr.5: 226122-226122 on Build GRCh38
rs59452511 Chr.5: 225826-225826 on Build GRCh38
rs189243489 Chr.5: 225855-225855 on Build GRCh38
rs143421811 Chr.5: 226182-226182 on Build GRCh38
rs768226227 Chr.5: 226059-226059 on Build GRCh38
rs553257776 Chr.5: 225988-225988 on Build GRCh38
rs569384870 Chr.5: 225892-225892 on Build GRCh38
rs147444427 Chr.5: 225978-225978 on Build GRCh38
rs587782076 Chr.5: 225938-225938 on Build GRCh38
rs753165149 Chr.5: 225962-225997 on Build GRCh38
rs777770952 Chr.5: 225959-225959 on Build GRCh38
rs201091275 Chr.5: 226055-226055 on Build GRCh38
rs899368013 Chr.5: 226031-226031 on Build GRCh38
rs776193478 Chr.5: 225937-225937 on Build GRCh38
rs775074528 Chr.5: 225904-225904 on Build GRCh38
rs755259343 Chr.5: 225866-225866 on Build GRCh38
rs772352951 Chr.5: 225810-225810 on Build GRCh38
rs761909900 Chr.5: 226084-226084 on Build GRCh38
rs899402014 Chr.5: 225805-225805 on Build GRCh38

View Full Product Details