Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 5187633-5188139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS16
Entrez Gene ID
170690
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 16
Gene Aliases
ADAMTS16s
UniGene
Hs.661915
Gene Chromosome Location
Chr.5: 5140330-5320304 on Build GRCh38
Transcripts
NM_139056.3 NR_136935.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4457031 c.1030C>T p.L344F Chr.5: 5187791-5187791 on Build GRCh38
6593949 c.1000A>C p.N334H Chr.5: 5187761-5187761 on Build GRCh38
3366090 c.1025T>A p.L342Q Chr.5: 5187786-5187786 on Build GRCh38
1634254 c.1007A>G p.N336S Chr.5: 5187768-5187768 on Build GRCh38
7137180 c.1031T>G p.L344R Chr.5: 5187792-5187792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374081297 Chr.5: 5187796-5187796 on Build GRCh38
rs747772414 Chr.5: 5187688-5187688 on Build GRCh38
rs948249229 Chr.5: 5187944-5187944 on Build GRCh38
rs1045611368 Chr.5: 5187998-5187999 on Build GRCh38
rs751510818 Chr.5: 5187752-5187752 on Build GRCh38
rs892220218 Chr.5: 5187722-5187722 on Build GRCh38
rs992239665 Chr.5: 5187895-5187895 on Build GRCh38
rs1863972 Chr.5: 5187663-5187663 on Build GRCh38
rs941755670 Chr.5: 5187893-5187893 on Build GRCh38
rs748904944 Chr.5: 5187809-5187809 on Build GRCh38
rs776005419 Chr.5: 5187711-5187711 on Build GRCh38
rs772999417 Chr.5: 5187703-5187703 on Build GRCh38
rs548006328 Chr.5: 5188067-5188067 on Build GRCh38
rs559685203 Chr.5: 5188058-5188058 on Build GRCh38
rs533622348 Chr.5: 5187976-5187976 on Build GRCh38
rs368399302 Chr.5: 5187801-5187801 on Build GRCh38
rs556694788 Chr.5: 5188107-5188107 on Build GRCh38
rs527277752 Chr.5: 5188084-5188084 on Build GRCh38
rs758161459 Chr.5: 5187678-5187678 on Build GRCh38
rs780828850 Chr.5: 5187800-5187800 on Build GRCh38
rs764998600 Chr.5: 5187675-5187675 on Build GRCh38
rs761906893 Chr.5: 5187736-5187736 on Build GRCh38
rs1010690849 Chr.5: 5188015-5188015 on Build GRCh38
rs373623862 Chr.5: 5187682-5187682 on Build GRCh38
rs151336838 Chr.5: 5187933-5187933 on Build GRCh38
rs1045350535 Chr.5: 5187681-5187681 on Build GRCh38
rs777156113 Chr.5: 5187839-5187839 on Build GRCh38
rs781143123 Chr.5: 5187977-5187977 on Build GRCh38
rs536885017 Chr.5: 5188094-5188094 on Build GRCh38
rs747481430 Chr.5: 5187830-5187830 on Build GRCh38
rs1030077636 Chr.5: 5187757-5187757 on Build GRCh38
rs936877228 Chr.5: 5187999-5187999 on Build GRCh38
rs761818824 Chr.5: 5187857-5187857 on Build GRCh38
rs1037509615 Chr.5: 5187912-5187912 on Build GRCh38
rs72647744 Chr.5: 5187859-5187859 on Build GRCh38
rs750187543 Chr.5: 5187677-5187677 on Build GRCh38
rs997170019 Chr.5: 5188037-5188037 on Build GRCh38
rs937156710 Chr.5: 5187967-5187967 on Build GRCh38
rs767735605 Chr.5: 5188013-5188013 on Build GRCh38
rs749290483 Chr.5: 5187815-5187815 on Build GRCh38
rs144922795 Chr.5: 5187717-5187717 on Build GRCh38
rs753847493 Chr.5: 5188108-5188108 on Build GRCh38
rs777417436 Chr.5: 5187808-5187808 on Build GRCh38
rs755715181 Chr.5: 5187802-5187802 on Build GRCh38
rs776728292 Chr.5: 5187718-5187718 on Build GRCh38
rs964236250 Chr.5: 5188089-5188089 on Build GRCh38
rs1016477817 Chr.5: 5188104-5188104 on Build GRCh38
rs182144689 Chr.5: 5188038-5188038 on Build GRCh38
rs765463357 Chr.5: 5187739-5187739 on Build GRCh38
rs577270970 Chr.5: 5187662-5187662 on Build GRCh38
rs896700260 Chr.5: 5187674-5187674 on Build GRCh38
rs564779359 Chr.5: 5187943-5187943 on Build GRCh38
rs775319896 Chr.5: 5187713-5187714 on Build GRCh38
rs554888512 Chr.5: 5187816-5187816 on Build GRCh38
rs769106091 Chr.5: 5187835-5187835 on Build GRCh38
rs751466415 Chr.5: 5187684-5187684 on Build GRCh38
rs750618810 Chr.5: 5187741-5187741 on Build GRCh38
rs373154678 Chr.5: 5188055-5188055 on Build GRCh38
rs772326555 Chr.5: 5187707-5187707 on Build GRCh38
rs531176405 Chr.5: 5187867-5187867 on Build GRCh38
rs370393357 Chr.5: 5187998-5187998 on Build GRCh38
rs545801213 Chr.5: 5187714-5187714 on Build GRCh38
rs78955888 Chr.5: 5188027-5188027 on Build GRCh38
rs774437548 Chr.5: 5187871-5187871 on Build GRCh38
rs763753109 Chr.5: 5187854-5187854 on Build GRCh38
rs769619029 Chr.5: 5187689-5187689 on Build GRCh38
rs371621693 Chr.5: 5187747-5187747 on Build GRCh38
rs1056563832 Chr.5: 5187671-5187671 on Build GRCh38
rs777313593 Chr.5: 5187699-5187699 on Build GRCh38
rs780916764 Chr.5: 5187687-5187687 on Build GRCh38
rs986349361 Chr.5: 5187841-5187841 on Build GRCh38
rs189830925 Chr.5: 5187787-5187787 on Build GRCh38
rs201553454 Chr.5: 5187849-5187852 on Build GRCh38
rs933015902 Chr.5: 5187844-5187844 on Build GRCh38

View Full Product Details