Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 7706604-7707119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY2
Entrez Gene ID
108
Gene Name
adenylate cyclase 2
Gene Aliases
AC2, HBAC2
UniGene
Hs.481545
Gene Chromosome Location
Chr.5: 7396230-7830081 on Build GRCh38
Transcripts
NM_020546.2 XM_011513942.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366162 c.571A>T p.K191* Chr.5: 7706745-7706745 on Build GRCh38
5756697 c.608G>A p.R203H Chr.5: 7706782-7706782 on Build GRCh38
739188 c.1211A>G p.D404G Chr.5: 7706845-7706845 on Build GRCh38
739189 c.1157T>C p.V386A Chr.5: 7706791-7706791 on Build GRCh38
6104735 c.1140C>T p.I380I Chr.5: 7706774-7706774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769705353 Chr.5: 7706696-7706696 on Build GRCh38
rs917944109 Chr.5: 7706671-7706671 on Build GRCh38
rs72710500 Chr.5: 7706909-7706909 on Build GRCh38
rs10475387 Chr.5: 7707065-7707065 on Build GRCh38
rs923823690 Chr.5: 7707041-7707041 on Build GRCh38
rs75833689 Chr.5: 7706914-7706914 on Build GRCh38
rs765849740 Chr.5: 7706718-7706718 on Build GRCh38
rs758927191 Chr.5: 7706954-7706954 on Build GRCh38
rs762581134 Chr.5: 7706711-7706711 on Build GRCh38
rs759196362 Chr.5: 7706885-7706885 on Build GRCh38
rs781370542 Chr.5: 7706920-7706920 on Build GRCh38
rs540480423 Chr.5: 7706967-7706967 on Build GRCh38
rs1005091141 Chr.5: 7707010-7707010 on Build GRCh38
rs1027129200 Chr.5: 7707007-7707007 on Build GRCh38
rs773797958 Chr.5: 7706878-7706878 on Build GRCh38
rs779534602 Chr.5: 7706748-7706748 on Build GRCh38
rs114107837 Chr.5: 7706720-7706720 on Build GRCh38
rs780627502 Chr.5: 7706742-7706742 on Build GRCh38
rs927321666 Chr.5: 7706771-7706771 on Build GRCh38
rs923720503 Chr.5: 7706861-7706861 on Build GRCh38
rs866258272 Chr.5: 7706752-7706752 on Build GRCh38
rs75986905 Chr.5: 7706876-7706876 on Build GRCh38
rs189747945 Chr.5: 7706783-7706783 on Build GRCh38
rs958711199 Chr.5: 7707048-7707048 on Build GRCh38
rs765490863 Chr.5: 7706925-7706925 on Build GRCh38
rs773017728 Chr.5: 7706704-7706704 on Build GRCh38
rs1024928713 Chr.5: 7707037-7707037 on Build GRCh38
rs377206615 Chr.5: 7706731-7706731 on Build GRCh38
rs751250778 Chr.5: 7706719-7706719 on Build GRCh38
rs373126277 Chr.5: 7707089-7707089 on Build GRCh38
rs970527679 Chr.5: 7707066-7707066 on Build GRCh38
rs531238561 Chr.5: 7707084-7707084 on Build GRCh38
rs897413310 Chr.5: 7706969-7706969 on Build GRCh38
rs771347600 Chr.5: 7706897-7706897 on Build GRCh38
rs576380123 Chr.5: 7706808-7706808 on Build GRCh38
rs940316445 Chr.5: 7707094-7707094 on Build GRCh38
rs936617861 Chr.5: 7707055-7707055 on Build GRCh38
rs1042737629 Chr.5: 7706717-7706717 on Build GRCh38
rs978122491 Chr.5: 7707039-7707039 on Build GRCh38
rs879015990 Chr.5: 7706722-7706722 on Build GRCh38
rs549115658 Chr.5: 7707038-7707038 on Build GRCh38
rs990684176 Chr.5: 7707049-7707049 on Build GRCh38
rs900227893 Chr.5: 7706786-7706786 on Build GRCh38
rs35209447 Chr.5: 7706789-7706789 on Build GRCh38
rs745750158 Chr.5: 7706668-7706668 on Build GRCh38
rs750875345 Chr.5: 7706767-7706767 on Build GRCh38
rs750972768 Chr.5: 7706932-7706932 on Build GRCh38
rs773778005 Chr.5: 7707083-7707083 on Build GRCh38
rs758592010 Chr.5: 7706774-7706774 on Build GRCh38
rs977867373 Chr.5: 7706798-7706798 on Build GRCh38
rs775804868 Chr.5: 7707095-7707095 on Build GRCh38
rs762765611 Chr.5: 7706834-7706834 on Build GRCh38
rs188317287 Chr.5: 7707031-7707031 on Build GRCh38
rs890154474 Chr.5: 7706903-7706903 on Build GRCh38
rs574535301 Chr.5: 7706638-7706638 on Build GRCh38
rs138459626 Chr.5: 7706641-7706641 on Build GRCh38
rs574326488 Chr.5: 7706643-7706643 on Build GRCh38
rs764461989 Chr.5: 7706729-7706729 on Build GRCh38
rs1051554389 Chr.5: 7707088-7707088 on Build GRCh38
rs573846891 Chr.5: 7706782-7706782 on Build GRCh38
rs781137779 Chr.5: 7706784-7706784 on Build GRCh38
rs10475386 Chr.5: 7707032-7707032 on Build GRCh38
rs112066650 Chr.5: 7706976-7706976 on Build GRCh38
rs758714787 Chr.5: 7706684-7706684 on Build GRCh38
rs773249308 Chr.5: 7706816-7706816 on Build GRCh38
rs990758079 Chr.5: 7706651-7706651 on Build GRCh38
rs535446509 Chr.5: 7707090-7707090 on Build GRCh38

View Full Product Details