The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030615.2 | NP_085118.2 | 8 - 9 | 1251 | 70 |
XM_011535802.2 | XP_011534104.1 | 10 - 11 | 1358 | 70 | |
GenBank mRNA | AB012722.2 | - | 8 - 9 | 1251 | 70 |
AK289488.1 | - | 10 - 11 | 1504 | 70 |