Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CYP2C8
- Entrez Gene ID
-
1558
- Gene Name
- cytochrome P450 family 2 subfamily C member 8
- Gene Aliases
-
CPC8, CYPIIC8, MP-12/MP-20
- Chromosome Location
-
Chr.10: 95036772 - 95069512 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M17398.1 |
- |
7 - 8 |
1148 |
108 |
View Full Product Details