The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018904.2 | NP_061727.1 | 1 - 1 | 602 | 120 |
NM_031865.1 | NP_114071.1 | 1 - 1 | 602 | 120 | |
GenBank mRNA | AB208924.1 | - | 1 - 1 | 663 | 120 |
AF152309.1 | - | 1 - 1 | 602 | 120 | |
AF152478.1 | - | 1 - 1 | 602 | 120 |