Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41917229-41917734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf51
Entrez Gene ID
285636
Gene Name
chromosome 5 open reading frame 51
Gene Aliases
-
UniGene
Hs.29899
Gene Chromosome Location
Chr.5: 41904313-41921636 on Build GRCh38
Transcripts
NM_175921.5 XM_005248289.4 XM_011514032.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3342387 c.856T>G p.L286V Chr.5: 41917270-41917270 on Build GRCh38
3828099 c.864C>A p.F288L Chr.5: 41917278-41917278 on Build GRCh38
188287 c.840G>A p.T280T Chr.5: 41917254-41917254 on Build GRCh38
6513033 c.823C>A p.Q275K Chr.5: 41917237-41917237 on Build GRCh38
243390 c.854T>G p.I285S Chr.5: 41917268-41917268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75553974 Chr.5: 41917603-41917603 on Build GRCh38
rs529880530 Chr.5: 41917629-41917629 on Build GRCh38
rs1012617759 Chr.5: 41917379-41917379 on Build GRCh38
rs145499229 Chr.5: 41917467-41917467 on Build GRCh38
rs200220094 Chr.5: 41917601-41917602 on Build GRCh38
rs1000673912 Chr.5: 41917620-41917620 on Build GRCh38
rs111486286 Chr.5: 41917560-41917560 on Build GRCh38
rs927033041 Chr.5: 41917468-41917468 on Build GRCh38
rs749715864 Chr.5: 41917348-41917348 on Build GRCh38
rs1028779657 Chr.5: 41917602-41917602 on Build GRCh38
rs575898205 Chr.5: 41917660-41917660 on Build GRCh38
rs201732963 Chr.5: 41917599-41917600 on Build GRCh38
rs989450204 Chr.5: 41917341-41917341 on Build GRCh38
rs1057375494 Chr.5: 41917289-41917289 on Build GRCh38
rs1015919871 Chr.5: 41917385-41917385 on Build GRCh38
rs142594823 Chr.5: 41917653-41917653 on Build GRCh38
rs539312466 Chr.5: 41917593-41917593 on Build GRCh38
rs745924482 Chr.5: 41917322-41917322 on Build GRCh38
rs988800319 Chr.5: 41917645-41917645 on Build GRCh38
rs975422350 Chr.5: 41917584-41917584 on Build GRCh38
rs780140105 Chr.5: 41917514-41917514 on Build GRCh38
rs11405992 Chr.5: 41917602-41917603 on Build GRCh38
rs144285732 Chr.5: 41917288-41917288 on Build GRCh38
rs374722484 Chr.5: 41917320-41917320 on Build GRCh38
rs963165952 Chr.5: 41917419-41917419 on Build GRCh38
rs889900786 Chr.5: 41917606-41917606 on Build GRCh38
rs763382482 Chr.5: 41917636-41917636 on Build GRCh38
rs548610582 Chr.5: 41917621-41917621 on Build GRCh38
rs971602259 Chr.5: 41917690-41917690 on Build GRCh38
rs954662642 Chr.5: 41917498-41917498 on Build GRCh38
rs542617961 Chr.5: 41917670-41917670 on Build GRCh38
rs930379257 Chr.5: 41917504-41917504 on Build GRCh38
rs772461387 Chr.5: 41917296-41917308 on Build GRCh38
rs950210919 Chr.5: 41917367-41917367 on Build GRCh38
rs977868451 Chr.5: 41917631-41917631 on Build GRCh38
rs375491494 Chr.5: 41917611-41917611 on Build GRCh38
rs1044550185 Chr.5: 41917408-41917408 on Build GRCh38
rs545770917 Chr.5: 41917597-41917599 on Build GRCh38
rs886094699 Chr.5: 41917549-41917549 on Build GRCh38
rs768911100 Chr.5: 41917350-41917350 on Build GRCh38
rs72746996 Chr.5: 41917550-41917550 on Build GRCh38
rs757148532 Chr.5: 41917309-41917309 on Build GRCh38
rs894753183 Chr.5: 41917704-41917705 on Build GRCh38
rs111784175 Chr.5: 41917534-41917534 on Build GRCh38
rs758118598 Chr.5: 41917275-41917275 on Build GRCh38
rs34573251 Chr.5: 41917603-41917604 on Build GRCh38
rs770042531 Chr.5: 41917324-41917324 on Build GRCh38
rs993647650 Chr.5: 41917428-41917428 on Build GRCh38
rs746926407 Chr.5: 41917306-41917306 on Build GRCh38
rs1033038177 Chr.5: 41917621-41917621 on Build GRCh38
rs775570594 Chr.5: 41917347-41917347 on Build GRCh38
rs938821511 Chr.5: 41917678-41917678 on Build GRCh38
rs368512609 Chr.5: 41917257-41917257 on Build GRCh38
rs776016005 Chr.5: 41917556-41917556 on Build GRCh38

View Full Product Details