Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 41917503-41918006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf51
Entrez Gene ID
285636
Gene Name
chromosome 5 open reading frame 51
Gene Aliases
-
UniGene
Hs.29899
Gene Chromosome Location
Chr.5: 41904313-41921636 on Build GRCh38
Transcripts
NM_175921.5 XM_005248289.4 XM_011514032.2

SNP Detail

SNP ID Chromosome Location
rs529880530 Chr.5: 41917629-41917629 on Build GRCh38
rs75553974 Chr.5: 41917603-41917603 on Build GRCh38
rs200220094 Chr.5: 41917601-41917602 on Build GRCh38
rs1000673912 Chr.5: 41917620-41917620 on Build GRCh38
rs111486286 Chr.5: 41917560-41917560 on Build GRCh38
rs989322510 Chr.5: 41917968-41917968 on Build GRCh38
rs1039976411 Chr.5: 41917826-41917826 on Build GRCh38
rs188522126 Chr.5: 41917851-41917851 on Build GRCh38
rs796984207 Chr.5: 41917737-41917737 on Build GRCh38
rs993264768 Chr.5: 41917832-41917834 on Build GRCh38
rs575898205 Chr.5: 41917660-41917660 on Build GRCh38
rs1028779657 Chr.5: 41917602-41917602 on Build GRCh38
rs201732963 Chr.5: 41917599-41917600 on Build GRCh38
rs772703053 Chr.5: 41917914-41917914 on Build GRCh38
rs142594823 Chr.5: 41917653-41917653 on Build GRCh38
rs539312466 Chr.5: 41917593-41917593 on Build GRCh38
rs150984820 Chr.5: 41917916-41917916 on Build GRCh38
rs988800319 Chr.5: 41917645-41917645 on Build GRCh38
rs975422350 Chr.5: 41917584-41917584 on Build GRCh38
rs1024310509 Chr.5: 41917942-41917942 on Build GRCh38
rs11405992 Chr.5: 41917602-41917603 on Build GRCh38
rs901805654 Chr.5: 41917865-41917865 on Build GRCh38
rs540359754 Chr.5: 41917797-41917797 on Build GRCh38
rs931890769 Chr.5: 41917880-41917880 on Build GRCh38
rs889900786 Chr.5: 41917606-41917606 on Build GRCh38
rs763382482 Chr.5: 41917636-41917636 on Build GRCh38
rs548610582 Chr.5: 41917621-41917621 on Build GRCh38
rs903678021 Chr.5: 41917890-41917890 on Build GRCh38
rs971602259 Chr.5: 41917690-41917690 on Build GRCh38
rs183730203 Chr.5: 41917739-41917739 on Build GRCh38
rs542617961 Chr.5: 41917670-41917670 on Build GRCh38
rs930441261 Chr.5: 41917767-41917767 on Build GRCh38
rs977868451 Chr.5: 41917631-41917631 on Build GRCh38
rs375491494 Chr.5: 41917611-41917611 on Build GRCh38
rs545770917 Chr.5: 41917597-41917599 on Build GRCh38
rs886094699 Chr.5: 41917549-41917549 on Build GRCh38
rs72746996 Chr.5: 41917550-41917550 on Build GRCh38
rs1000680739 Chr.5: 41917894-41917894 on Build GRCh38
rs554564083 Chr.5: 41917733-41917733 on Build GRCh38
rs894753183 Chr.5: 41917704-41917705 on Build GRCh38
rs111784175 Chr.5: 41917534-41917534 on Build GRCh38
rs890196782 Chr.5: 41917883-41917887 on Build GRCh38
rs984931986 Chr.5: 41917774-41917774 on Build GRCh38
rs34573251 Chr.5: 41917603-41917604 on Build GRCh38
rs907586621 Chr.5: 41917810-41917810 on Build GRCh38
rs1033524019 Chr.5: 41917908-41917908 on Build GRCh38
rs891845205 Chr.5: 41917911-41917911 on Build GRCh38
rs1033038177 Chr.5: 41917621-41917621 on Build GRCh38
rs940355366 Chr.5: 41917815-41917815 on Build GRCh38
rs1050249688 Chr.5: 41917889-41917889 on Build GRCh38
rs927439706 Chr.5: 41917765-41917765 on Build GRCh38
rs938821511 Chr.5: 41917678-41917678 on Build GRCh38
rs776016005 Chr.5: 41917556-41917556 on Build GRCh38

View Full Product Details