The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032149.2 | NP_115525.2 | 2 - 3 | 485 | 82 |
XM_011532315.1 | XP_011530617.1 | 2 - 3 | 465 | 82 | |
GenBank mRNA | AK093085.1 | - | 2 - 3 | 477 | 82 |
AK223345.1 | - | 2 - 3 | 472 | 82 | |
AK314711.1 | - | 2 - 3 | 468 | 82 | |
AL136838.1 | - | 2 - 3 | 446 | 82 | |
BC074759.2 | - | 1 - 2 | 162 | 82 | |
CR533542.1 | - | 1 - 2 | 123 | 82 | |
DB051883.1 | - | 2 - 3 | 485 | 82 |