The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032151.4 | NP_115527.3 | 1 - 2 | 103 | 126 |
GenBank mRNA | AK098495.1 | - | 1 - 2 | 75 | 126 |
AL136721.1 | - | 1 - 2 | 103 | 126 | |
BC054021.1 | - | 1 - 2 | 106 | 126 |